Merck
  • A fluorescence high throughput screening method for the detection of reactive electrophiles as potential skin sensitizers.
A fluorescence high throughput screening method for the detection of reactive electrophiles as potential skin sensitizers.
Toxicology and applied pharmacology (2015-10-13)
Cristina Avonto, Amar G Chittiboyina, Diego Rua, Ikhlas A Khan
ABSTRACT
Skin sensitization is an important toxicological end-point in the risk assessment of chemical allergens. Because of the complexity of the biological mechanisms associated with skin sensitization, integrated approaches combining different chemical, biological and in silico methods are recommended to replace conventional animal tests. Chemical methods are intended to characterize the potential of a sensitizer to induce earlier molecular initiating events. The presence of an electrophilic mechanistic domain is considered one of the essential chemical features to covalently bind to the biological target and induce further haptenation processes. Current in chemico assays rely on the quantification of unreacted model nucleophiles after incubation with the candidate sensitizer. In the current study, a new fluorescence-based method, 'HTS-DCYA assay', is proposed. The assay aims at the identification of reactive electrophiles based on their chemical reactivity toward a model fluorescent thiol. The reaction workflow enabled the development of a High Throughput Screening (HTS) method to directly quantify the reaction adducts. The reaction conditions have been optimized to minimize solubility issues, oxidative side reactions and increase the throughput of the assay while minimizing the reaction time, which are common issues with existing methods. Thirty-six chemicals previously classified with LLNA, DPRA or KeratinoSens™ were tested as a proof of concept. Preliminary results gave an estimated 82% accuracy, 78% sensitivity, 90% specificity, comparable to other in chemico methods such as Cys-DPRA. In addition to validated chemicals, six natural products were analyzed and a prediction of their sensitization potential is presented for the first time.
Sigma-Aldrich
Alantolactone, ≥98% (HPLC)
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Diethyl maleate, 97%
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
2,4-Heptadienal, technical grade, 90%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG
Sigma-Aldrich
Diethyl maleate, ≥96%, FG
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%