Merck
Cytochrome p450-mediated metabolic activation of diosbulbin B.
Drug metabolism and disposition: the biological fate of chemicals (2014-07-16)
Dongju Lin, Chunyan Li, Ying Peng, Huiyuan Gao, Jiang Zheng
ABSTRACT
Diosbulbin B (DIOB), a furan-containing diterpenoid lactone, is the most abundant component of Dioscorea bulbifera L. (DB), a traditional Chinese medicine herb. Administration of purified DIOB or DB extracts has been reported to cause liver injury in animals. The mechanisms of DIOB-induced hepatotoxicity remain unknown. The major objective of this study was to identify reactive metabolites of DIOB. A DIOB-derived cis-enedial was trapped by N-acetyl lysine (NAL) and glutathione (GSH) or N-acetyl cysteine (NAC) in rat and human liver microsomal incubation systems after exposure to DIOB. Four metabolites (M1-M4) associated with GSH were detected by liquid chromatography coupled to tandem mass spectrometry. Apparently, M1 was derived from both NAL and GSH. M2 and M3 resulted from the reaction of GSH without the involvement of NAL. Two molecules of GSH participated in the formation of M4. M2 and M3 were also detected in bile and urine of rats given DIOB. M5, a DIOB-derived NAC/NAL conjugate, was detected in microsomal incubations with DIOB fortified with NAC and NAL as trapping agents. Biomimetic M1-M5 were prepared by oxidation of DIOB with Oxone for metabolite identification. Microsomal incubation study demonstrated that ketoconazole inhibited the production of the enedial in a concentration-dependent manner, and CYP3A4 was found to be the enzyme responsible for the metabolic activation of DIOB. The metabolism study facilitates the understanding of the role of bioactivation of DIOB in its hepatotoxicity.
Product Description
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Xanthotoxin, analytical standard
Sigma-Aldrich
Cytochrome P450 human, 1A2 Isozyme Microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 1A1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 4F3B isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)
Sigma-Aldrich
Cytochrome P450 human, 3A4 isozyme microsomes, with P450 reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Supelco
8-Methoxypsoralen, analytical standard
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytochrome P450 human, 2E1 isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Quinidine, anhydrous
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cytochrome P450 human, 1B1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nα-Acetyl-L-lysine
Disulfiram, European Pharmacopoeia (EP) Reference Standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material