Skip to Content
Merck
All Photos(1)

Key Documents

K0600000

Ketoconazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-cis-1-Acetyl-4-(4-[(2-[2,4-dichlorophenyl]-2-[1H-imidazol-1-ylmethyl]-1,3-dioxolan-4-yl)-methoxy]phenyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N4O4
CAS Number:
Molecular Weight:
531.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketoconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)N1CCN(CC1)c2ccc(OC[C@H]3CO[C@@](Cn4ccnc4)(O3)c5ccc(Cl)cc5Cl)cc2

InChI

1S/C26H28Cl2N4O4/c1-19(33)31-10-12-32(13-11-31)21-3-5-22(6-4-21)34-15-23-16-35-26(36-23,17-30-9-8-29-18-30)24-7-2-20(27)14-25(24)28/h2-9,14,18,23H,10-13,15-17H2,1H3/t23-,26-/m0/s1

InChI key

XMAYWYJOQHXEEK-OZXSUGGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

CYP3A4 inhibitor

Biochem/physiol Actions

Antifungal agent

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Halama et al.
Clinical pharmacology and therapeutics, 93(6), 564-571 (2013-03-21)
The objective of the study was to establish an in vivo method for assessing cytochrome P450 3A (CYP3A) activity using therapeutically inert nanogram doses of midazolam. We administered four escalating single doses of oral midazolam (0.0001-3 mg) to 12 healthy participants
Masato Ohbuchi et al.
Antimicrobial agents and chemotherapy, 57(3), 1332-1338 (2013-01-01)
The antifungal efficacy of voriconazole (VRC) differs among host species, with potent efficacy in humans but less in rodents. We investigated the possible involvement of pregnane X receptor (PXR) and constitutive androstane receptor (CAR) in the species-specific efficacy of VRC
K-H Shin et al.
Clinical pharmacology and therapeutics, 94(5), 601-609 (2013-06-21)
This study aimed to evaluate endogenous metabolic markers of hepatic cytochrome P450 (CYP)3A activity in healthy subjects using a metabolomics approach. Twenty-four subjects received the following medication during the following three study periods: 1 mg of i.v. midazolam alone (control
Jan Faergemann et al.
Expert opinion on pharmacotherapy, 8(9), 1365-1371 (2007-06-15)
Seborrhoeic dermatitis (SD) is a chronic, inflammatory skin disorder, affecting areas of the head and body where sebaceous glands are most prominent and active. The disorder commonly affects hair-bearing areas of the head, including the scalp. Involvement on the face
Hao Li et al.
The Journal of biological chemistry, 288(19), 13655-13668 (2013-03-26)
Ketoconazole binds to and antagonizes pregnane X receptor (PXR) activation. Yeast high throughput screens of PXR mutants define a unique region for ketoconazole binding. Ketoconazole genetically interacts with specific PXR surface residues. A yeast-based genetic method to discover novel nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service