Merck
  • Electrocatalytic reduction of CO2 with palladium bis-N-heterocyclic carbene pincer complexes.
Electrocatalytic reduction of CO2 with palladium bis-N-heterocyclic carbene pincer complexes.
Inorganic chemistry (2014-10-23)
Jeffrey A Therrien, Michael O Wolf, Brian O Patrick
ABSTRACT
A series of pyridine- and lutidine-linked bis-N-heterocyclic carbene (NHC) palladium pincer complexes were electrochemically characterized and screened for CO2 reduction capability with 2,2,2-trifluoroethanol, acetic acid, or 2,2,2-trifluoroacetic acid (TFA) as proton sources. The lutidine-linked pincer complexes electrocatalytically reduce CO2 to CO at potentials as low as -1.6 V versus Ag/AgNO3 in the presence of TFA. The one-electron reduction of these complexes is shown to be chemically reversible, yielding a monometallic species, with density functional theory studies indicating charge storage on the redox-active ligand, thus addressing a major source of deactivation in earlier triphosphine electrocatalysts.
Sigma-Aldrich
Potassium hexafluorophosphate, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Potassium hexafluorophosphate, 99.5% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Magnesium perchlorate, puriss., free-flowing powder, ≥99.0% (calc. based on dry substance, KT)
Sigma-Aldrich
Magnesium perchlorate, puriss. p.a., drying agent, ACS reagent, ≥98.0% (calc. based on dry substance, KT)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Magnesium perchlorate, ACS reagent
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%