Skip to Content
Merck
All Photos(2)

Key Documents

D158550

Sigma-Aldrich

N,N-Dimethylformamide

ReagentPlus®, ≥99%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

dilution

(for general lab use)

impurities

≤0.1% Water (Karl Fischer)

refractive index

n20/D 1.430 (lit.)

pH

6.7

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide (DMF) is a polar organic solvent with a wide range of industrial applications.[1] It is a precursor for numerous reactions such as formylation, aminocarbonylation, amination, amidation and cyanation.[2] Along with phosphorus oxychloride, it forms Vilsmeier reagent used in the formylation of reactive aromatic and heteroaromatic substrates.[3] Dielectric relaxation experiments on DMF have been conducted between 238.15 K and 338.15K.[4] The thermophysical properties of the molecular interactions between DMF and the ionic liquids, ammonium salts and imidazolium salts have been studied.[5] The characteristics of solute-solvent interactions between ammonium nitrate and DMF have been investigated.[6]

Application

N,N-Dimethylformamide has been used in the to prepare pure polyurethane as well as nanocomposite solutions.
It may be used in the following processes:
  • Synthesis of gold nanoparticles.[7]
  • Capillary electrophoresis.[8]
  • Regioselective synthesis of N,N-dimethylaminopyridzines.[9]
  • Synthesis of urea-N,N-dimethylformamide, a 3:1 solvate with urea.[10]
Solvent for many hydrophobic organic compounds.

Other Notes

A Greener alternative is available for many DMF applications, Cyrene (807796)

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capillary electrophoresis in N,N-dimethylformamide.
Porras SP and Kenndler E.
Electrophoresis, 26(17), 3279-3291 (2005)
Urea-N,N-dimethylformamide (3/1).
Fernandes P, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(12), 4861-4861 (2007)
Reaction of chloropyridazines with N,N -dimethylformamide.
Lee WS, et al.
Journal of Heterocyclic Chemistry, 37(6), 1591-1591 (2000)
N,N-dimethylformamide as a reaction medium for metal nanoparticle synthesis.
Pastoriza-Santos I and Liz-Marzan LM.
Advances in Functional Materials, 19(5), 679-688 (2009)
N,N-Dimethylformamide: much more than a solvent.
Muzart J.
Tetrahedron, 65(40), 8313-8323 (2009)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service