Skip to Content
Merck
All Photos(3)

Documents

34998

Sigma-Aldrich

Acetonitrile

≥99.9%, HPLC Plus, suitable for IR spectroscopy, HPLC

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
41116105
eCl@ss:
39031501
PubChem Substance ID:
NACRES:
NA.03

product name

Acetonitrile, HPLC Plus, ≥99.9%

grade

HPLC Plus

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

baseline drift measuring range

≤12 mAU (210 nm)

technique(s)

HPLC: suitable
IR spectroscopy: suitable

impurities

≤0.0001% free alkali (as NH3)
≤0.0002% non-volatile matter
≤0.001% free acid (as CH3COOH)
≤0.01% water (Karl Fischer)
≤0.5 ppb fluorescence (quinine) at 254 nm
≤0.5 ppb fluorescence (quinine) at 365 nm

halogenated residue

10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

HPLC-gradient

≤0.2 mAU at 254 nm
≤1 mAU at 210 nm

λ

H2O reference

UV absorption

λ: 195 nm Amax: ≤0.10
λ: 200 nm Amax: ≤0.02
λ: 228 nm Amax: ≤0.005
λ: 235 nm Amax: ≤0.0044
λ: 250 nm Amax: ≤0.0044
λ: 400 nm Amax: ≤0.0044

application(s)

food and beverages

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile (MeCN) is widely employed as a solvent in various studies and has high dielectric constant (37.5). Various purification procedures to obtain different grades of acetonitrile for use in different studies (polarography, spectroscopy, etc.) have been reported by many researchers.

Application

Acetonitrile has been used to compose the gradient system for the HPLC detection of diphenylmethane diisocyanate, a polymeric degradation product.
New Quality UPLC Plus Acetonitrile is suitable for all applications including:
Mass spectrometry, HPLC, TLC, GPC, UV, IR, GC, Pesticide Residue Analysis, and RNA/DNA Synthesis and Purification.

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Important notice
  • The article number 34998-4X2.5L will be discontinued. Please order the single bottle 34998-2.5L which is physically identical with the same exact specifications.
  • The article number 34998-6X1L will be discontinued. Please order the single bottle 34998-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H Haugen et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 73(2), 229-237 (2005-03-10)
This article deals with enzyme-induced biodegradation behavior of thermoplastic polyether-urethane (TPU). Porous scaffolds were processed by a new foaming method applied in hot pressing and injection molding. The scaffolds were subsequently gamma sterilized. The samples were incubated with cholesterol esterase
Alexander David Ardianrama et al.
Journal of colloid and interface science, 552, 485-493 (2019-06-04)
Although triangular silver (Ag) nanoplates are intrinsically unstable, this characteristic has been taken advantage of in the development of a novel sensing platform. However, most of these applications have relied on halide ions as etchants. In the current work, we
Purification of acetonitrile.
Walter M and Ramaley L.
Analytical Chemistry, 45(1), 165-166 (1973)
Timo Glatter et al.
Molecular systems biology, 5, 237-237 (2009-01-22)
Protein complexes represent major functional units for the execution of biological processes. Systematic affinity purification coupled with mass spectrometry (AP-MS) yielded a wealth of information on the compendium of protein complexes expressed in Saccharomyces cerevisiae. However, global AP-MS analysis of
Hélène Hall et al.
Brain : a journal of neurology, 137(Pt 9), 2493-2508 (2014-07-27)
The neuropathological substrate of dementia in patients with Parkinson's disease is still under debate, particularly in patients with insufficient alternate neuropathology for other degenerative dementias. In patients with pure Lewy body Parkinson's disease, previous post-mortem studies have shown that dopaminergic

Protocols

HPLC Analysis of DMB-Labeled Sialic Acids on Ascentis ® Express RP-Amide: Comparison of Biosimilars to Reference Materials

HPLC Analysis of Vitamins B1, B6 and B12 using Ascentis® Express RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service