Merck
  • Is the full potential of the biopharmaceutics classification system reached?
Is the full potential of the biopharmaceutics classification system reached?
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2013-10-01)
Christel A S Bergström, Sara B E Andersson, Jonas H Fagerberg, Gert Ragnarsson, Anders Lindahl
ABSTRACT
In this paper we analyse how the biopharmaceutics classification system (BCS) has been used to date. A survey of the literature resulted in a compilation of 242 compounds for which BCS classes were reported. Of these, 183 compounds had been reported to belong to one specific BCS class whereas 59 compounds had been assigned to multiple BCS classes in different papers. Interestingly, a majority of the BCS class 2 compounds had fraction absorbed (FA) values >85%, indicating that they were completely absorbed after oral administration. Solubility was computationally predicted at pH 6.8 for BCS class 2 compounds to explore the impact of the pH of the small intestine, where most of the absorption occurs, on the solubility. In addition, the solubilization capacity of lipid aggregates naturally present in the intestine was studied computationally and experimentally for a subset of 12 compounds. It was found that all acidic compounds with FA>85% were completely dissolved in the pH of the small intestine. Further, lipids at the concentration used in fasted state simulated intestinal fluid (FaSSIF) dissolved the complete dose given of the most lipophilic (logD6.5>3) compounds studied. Overall, biorelevant dissolution media (pure buffer of intestinal pH or FaSSIF) identified that for 20 of the 29 BCS class 2 compounds with FA>85% the complete dose given orally would be dissolved. These results indicate that a more relevant pH restriction for acids and/or dissolution medium with lipids present better forecast solubility-limited absorption in vivo than the presently used BCS solubility criterion. The analysis presented herein further strengthens the discussion on the requirement of more physiologically relevant dissolution media for the in vitro solubility classification performed to reach the full potential of the BCS.
Product Description
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
4-Aminophenyl sulfone, 97%
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Lansoprazole, ≥98% (TLC), powder
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Lansoprazole, European Pharmacopoeia (EP) Reference Standard
Lansoprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lansoprazole, United States Pharmacopeia (USP) Reference Standard
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Praziquantel for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rofecoxib, ≥98% (HPLC)
Supelco
Carbamazepine, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Dapsone, VETRANAL®, analytical standard
Supelco
Praziquantel, VETRANAL®, analytical standard
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O