Skip to Content
Merck
All Photos(1)

Key Documents

1356916

USP

Lansoprazole

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3N3O2S
CAS Number:
Molecular Weight:
369.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

lansoprazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1c(CS(=O)c2nc3ccccc3[nH]2)nccc1OCC(F)(F)F

InChI

1S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)

InChI key

MJIHNNLFOKEZEW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lansoprazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Lansoprazole Compounded Oral Suspension
  • Lansoprazole Delayed-Release Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Iwahi et al.
Antimicrobial agents and chemotherapy, 35(3), 490-496 (1991-03-01)
The activities of various types of antiulcer agents against Helicobacter pylori (formerly called Campylobacter pylori) strains were determined by an agar dilution method. Among the compounds tested, two benzimidazole proton pump inhibitors, lansoprazole (AG-1749) and omeprazole, were found to have
A modified high-performance liquid chromatographic method for the analysis of pantoprazole sodium in pharmaceutical dosage forms using lansoprazole as internal standard.
Ashour, Safwan, and Soulafa Omar.
Arabian Journal of Chemistry, 1-6 (2011)
Ken Haruma et al.
Internal medicine (Tokyo, Japan), 54(7), 695-701 (2015-04-04)
Although gastroesophageal reflux disease (GERD), a very common disorder worldwide, is considered to be a lifestyle disease, the pathogenic role of lifestyle factors and consequently the efficacy of lifestyle interventions, remains controversial. Lifestyle factors associated with GERD and the beneficial
Hiroto Miwa et al.
Journal of gastroenterology and hepatology, 30(5), 834-841 (2015-01-07)
Non-erosive reflux disease (NERD) and functional dyspepsia (FD) share some common features, and the appropriateness of managing these diseases independently has been questioned. A post-hoc subgroup analysis of the LEGEND study was performed to compare demographics and treatment response between
Yasunobu Yamashita et al.
PloS one, 9(5), e97419-e97419 (2014-05-23)
Lansoprazole is a potent anti-gastric ulcer drug that inhibits gastric proton pump activity. We identified a novel function for lansoprazole, as an inducer of anti-oxidative stress responses in the liver. Gastric administration of lansoprazole (10-100 mg/kg) to male Wistar rats

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service