Skip to Content
Merck
All Photos(3)

Documents

R7382

Sigma-Aldrich

Rifampicin

powder or crystals, suitable for plant cell culture, BioReagent

Synonym(s):

3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94
Beilstein:
5723476
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product name

Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals

product line

BioReagent

Assay

≥95% (HPLC)

form

powder or crystals

technique(s)

cell culture | plant: suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

application(s)

agriculture

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)c(\C=N\N5CCN(C)CC5)c(O)c4c3C2=O

InChI

1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1

InChI key

JQXXHWHPUNPDRT-WLSIYKJHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Rifampicin, a rifamycin, is active against mycobacteria. Rifamycin, is also an inhibitor of chloroplast RNA polymerase and may be used to study chloroplast-level DNA transcription in plants.

Biochem/physiol Actions

Inhibits the assembly of DNA and protein into mature virus particles.
Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Product is sensitive to light and moisture. Store under inert gas.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan Eberhard et al.
The Plant journal : for cell and molecular biology, 31(2), 149-160 (2002-07-18)
We performed a systematic investigation of the quantitative relationship between genome copy number, transcription, transcript abundance and synthesis of photosynthetic proteins in the chloroplast of the green algae Chlamydomonas reinhardtii grown either in mixotrophic or phototrophic conditions. The chloroplast gene
Igor Gurevich et al.
Biology open, 9(12) (2020-12-04)
Non-alcoholic fatty liver disease (NAFLD) affects 30-40% of adults and 10% of children in the US. About 20% of people with NAFLD develop non-alcoholic steatohepatitis (NASH), which may lead to cirrhosis and liver cancer, and is projected to be a
Sandra V Kik et al.
The Journal of infectious diseases, 211 Suppl 2, S58-S66 (2015-03-15)
The potential available market (PAM) for new diagnostics for tuberculosis that meet the specifications of the high-priority target product profiles (TPPs) is currently unknown. We estimated the PAM in 2020 in 4 high-burden countries (South Africa, Brazil, China, and India)
Daniela Baldoni et al.
Antimicrobial agents and chemotherapy, 53(3), 1142-1148 (2008-12-17)
We investigated the activity of linezolid, alone and in combination with rifampin (rifampicin), against a methicillin-resistant Staphylococcus aureus (MRSA) strain in vitro and in a guinea pig model of foreign-body infection. The MIC, minimal bactericidal concentration (MBC) in logarithmic phase
Rodney Dawson et al.
Lancet (London, England), 385(9979), 1738-1747 (2015-03-22)
New antituberculosis regimens are urgently needed to shorten tuberculosis treatment. Following on from favourable assessment in a 2 week study, we investigated a novel regimen for efficacy and safety in drug-susceptible and multidrug-resistant (MDR) tuberculosis during the first 8 weeks

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service