Merck
  • Micro-scale strategy to detect spermine and spermidine by MALDI-TOF MS in foods and identification of apoptosis-related proteins by nano-flow UPLC-MS/MS after treatment with spermine and spermidine.
Micro-scale strategy to detect spermine and spermidine by MALDI-TOF MS in foods and identification of apoptosis-related proteins by nano-flow UPLC-MS/MS after treatment with spermine and spermidine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-30)
Huai-Hsin Su, Lea-Yea Chuang, Wei-Lung Tseng, Chi-Yu Lu
ABSTRACT
Spermine and spermidine are multiple-nitrogen compounds found in many foods. Both compounds are essential for cell growth and human health. This study established a simple and fast method of detecting spermine and spermidine in food samples by matrix-assisted laser desorption/ionization combined with time-of-flight mass spectrometry (MALDI-TOF MS). After a simple sample preparation procedure, spermine and spermidine were directly detected by MALDI-TOF MS with no additional purification procedure. The calibration curves for spermine and spermidine ranged from 0.1 to 10 μg/mL. In intra- and inter-batch assays of three different concentrations of spermine and spermidine, all relative standard deviations and relative errors were below 18.9%. These experimental results confirmed the practicability and effectiveness of the proposed MALDI-TOF MS method for fast determination of spermine and spermidine in food samples. Furthermore, since spermine and spermidine have important roles in apoptosis, up-regulation and down-regulation of spermine and spermidine during apoptosis were analyzed. After treating NRK-52E cells with spermine and spermidine, the cells were lysed, and cell proteins were collected, and digested. Apoptosis-related proteins were then identified by tandem MS.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Mercaptobenzothiazole, 97%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M