Skip to Content
Merck

Skip To

72064

Sodium hydroxide solution

49-51% in water, eluent for IC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
NACRES:
NB.21
PubChem Substance ID:
UNSPSC Code:
12352316
MDL number:
Concentration:
49-51% in water
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

3 mmHg ( 37 °C)

Quality Segment

form

liquid

concentration

49-51% in water

technique(s)

ion chromatography: suitable

impurities

≤0.0005% total nitrogen (N), ≤0.01% ammonium hydroxide precipitate

pH

14.0

solubility

water: soluble (completely miscible)

anion traces

carbonate (as Na2CO3): ≤1000 mg/kg, chloride (Cl-): ≤100 mg/kg, phosphate (PO43-): ≤5 mg/kg, silicate (as SiO2): ≤10 mg/kg, sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤100 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤10 mg/kg, Zn: ≤5 mg/kg

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

General description

Sodium hydroxide solution is a strong alkaline solution which may be suitable to make dyes, drugs, cellophane, rayon and bleaches. It may also be suitable for petrochemical industry to remove impurities. This solution needs to be handled carefully, close proximity or ingestion could cause blindness, permanent scarring and sometimes can be fatal.

Application

Sodium hydroxide solution may be used to characterize the morphology of the produced colloids using UV-Visible spectroscopy and transmission electron microscopy. It may be used to measure specific heats using calorimeter. It may also be used to study the first stages of silver oxidation using voltammetry and ellipsometry.

Metrohm IC application note AN-P-055:Lactose residues in "lactose-free" milk.

Other Notes

Visit the IC Portal to learn more

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
415413S277005211
concentration

49-51% in water

concentration

48-52% (by HCl, titration), 50% in H2O

concentration

1.0 N

concentration

≥32%

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

form

liquid

form

liquid (viscous)

form

solution

form

solution

pH

14.0

pH

14

pH

-

pH

14

solubility

water: soluble (completely miscible)

solubility

water: soluble

solubility

-

solubility

-

impurities

≤0.0005% total nitrogen (N), ≤0.01% ammonium hydroxide precipitate

impurities

-

impurities

-

impurities

≤0.001% heavy metals (as Pb)


Still not finding the right product?

Explore all of our products under Sodium hydroxide solution


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters