Skip to Content
Merck
All Photos(3)

Key Documents

30620

Sigma-Aldrich

Sodium hydroxide

puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

≥98%

form

pellets

technique(s)

titration: suitable

impurities

≤0.0003% total nitrogen (N)
≤0.0005% heavy metals (as Pb)
≤1% Na2CO3

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

density

2.13 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
silicate (as SiO2): ≤10 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤1 mg/kg
Ca: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Hg: ≤0.05 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤2 mg/kg
Zn: ≤5 mg/kg

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide is an inorganic base. It participates in the oxidation of glycerol, catalyzed by Au/charcoal or Au/graphite. It also participates in solvent-free aldol condensation reaction.

Application

Sodium hydroxide may be employed for determination of specific surface area of colloidal silica, via titration. It may be employed for direct oxidation of sodium borohydride, which was investigated by cyclic and linear voltammetry, chronoamperometry and chronopotentiometry. It may be used to compose citrate buffer and borate buffers.
It may be used in the synthesis of following compounds:
  • alkylaryl amidosulfobetaines
  • (3 -[4-teroctylphenoxyethoxyethoxyethyl] dimethylammonio propane sulfonate (XOSB)
  • trisulfobetaines (TriSBn)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvio Carrettin et al.
Chemical communications (Cambridge, England), (7)(7), 696-697 (2002-07-18)
Glycerol is oxidised to glyceric acid with 100% selectivity using either 1% Au/charcoal or 1% Au/graphite catalyst under mild reaction conditions (60 degrees C, 3 h, water as solvent).
Determination of specific surface area of colloidal silica by titration with sodium hydroxide.
Sears Jr GW.
Analytical Chemistry, 28(12), 1981-1983 (1956)
Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
Kinetics of sodium borohydride direct oxidation and oxygen reduction in sodium hydroxide electrolyte: Part I. BH4- electro-oxidation on Au and Ag catalysts.
Chatenet M, et al.
Electrochimica Acta, 51(25), 5459-5467 (2006)
M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service