Skip to Content
Merck
All Photos(1)

Key Documents

S0650000

Simvastatin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H38O5
CAS Number:
Molecular Weight:
418.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

simvastatin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

127-132 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12[C@H](C[C@@H](C)C=C1C=C[C@H](C)[C@@H]2CC[C@@H]3C[C@@H](O)CC(=O)O3)OC(=O)C(C)(C)CC

InChI

1S/C25H38O5/c1-6-25(4,5)24(28)30-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-19-13-18(26)14-22(27)29-19/h7-8,11,15-16,18-21,23,26H,6,9-10,12-14H2,1-5H3/t15-,16-,18+,19+,20-,21-,23-/m0/s1

InChI key

RYMZZMVNJRMUDD-HGQWONQESA-N

Gene Information

human ... HMGCR(3156)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Simvastatin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Simvastatin is a specific inhibitor of HMG-CoA reductase and cholesterol lowering drug.
Simvastatin is a specific inhibitor of HMG-CoA reductase, the enzyme that catalyzes the conversion of HMG-CoA to mevalonate, an early step in cholesterol biosynthesis. It is used in the treatment of hypercholesterolemia, as it reduces levels of low-density lipoproteins and triglycerides, and raises high-density lipoprotein levels. Simvastatin is a lactone that is readily hydrolyzed in vivo to the corresponding β-hydroxyacid, and can be activated prior to use with NaOH in EtOH treatment. It is a synthetic analog of lovastatin (Cat. No. M2147).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veluchamy A Barathi et al.
Investigative ophthalmology & visual science, 55(1), 460-468 (2014-01-09)
To determine the effect of a statin (simvastatin) on the ultrastructure and function of the RPE, Bruch's membrane (BM), and photoreceptor interface in a high-fat atherogenic mouse model of thickened BM. Wild-type C57BL/6 mice (6-weeks old) were divided into three
Thijs van der Vaart et al.
The Lancet. Neurology, 12(11), 1076-1083 (2013-10-05)
Neurofibromatosis type 1 is a common genetic disorder characterised by neurocutaneous manifestations and cognitive and behavioural problems. Statins were shown to reduce analogous learning deficits in a mouse model of the disease, but a short-term trial in humans was inconclusive.
Kirill Tarasov et al.
The Journal of clinical endocrinology and metabolism, 99(1), E45-E52 (2013-11-19)
Coronary artery disease (CAD) is among the leading causes of mortality and morbidity worldwide. Traditional risk markers explain only a proportion of total cardiovascular risk. Thus, development and improvement of early diagnostic strategies and targeted initiation of preventive measures would
Heiner K Berthold et al.
PloS one, 8(3), e60095-e60095 (2013-04-02)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) is a secreted inhibitor of the low-density lipoprotein (LDL) receptor and an important regulator of LDL metabolism. Elevated PCSK9 levels have been associated with cardiovascular risk. The purpose of this study was to investigate
Y Zhu et al.
Cell death & disease, 4, e568-e568 (2013-04-06)
Although statins are known to inhibit proliferation and induce death in a number of cancer cell types, the mechanisms through which downregulation of the mevalonate (MVA) pathway activates death signaling remain poorly understood. Here we set out to unravel the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service