Merck
  • Regulatory T cell-derived adenosine induces dendritic cell migration through the Epac-Rap1 pathway.
Regulatory T cell-derived adenosine induces dendritic cell migration through the Epac-Rap1 pathway.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-18)
Sabine Ring, Anna Pushkarevskaya, Hansjörg Schild, Hans Christian Probst, Verena Jendrossek, Florian Wirsdörfer, Catherine Ledent, Simon Christopher Robson, Alexander H Enk, Karsten Mahnke
ABSTRACT
Dendritic cells (DC) are one target for immune suppression by regulatory T cells (Treg), because their interaction results in reduced T cell stimulatory capacity and secretion of inhibitory cytokines in DC. We show that DC in the presence of Treg are more mobile as compared with cocultures with conventional CD4(+) T cells and form DC-Treg aggregates within 2 h of culture. The migration of DC was specifically directed toward Treg, as Treg, but not CD4(+) T cells, attracted DC in Boyden chambers. Treg deficient for the ectonucleotidase CD39 were unable to attract DC. Likewise, addition of antagonists for A2A adenosine receptors abolished the formation of DC-Treg clusters, indicating a role for adenosine in guiding DC-Treg interactions. Analysis of the signal transduction events in DC after contact to Treg revealed increased levels of cAMP, followed by activation of Epac1 and the GTPase Rap1. Subsequently activated Rap1 localized to the subcortical actin cytoskeleton in DC, providing a means by which directed locomotion of DC toward Treg is facilitated. In aggregate, these data show that Treg degrade ATP to adenosine via CD39, attracting DC by activating Epac1-Rap1-dependent pathways. As a consequence, DC-Treg clusters are formed and DC are rendered less stimulatory. This adenosine-mediated attraction of DC may therefore act as one mechanism by which Treg regulate the induction of immune responses by DC.
Product Description
Supelco
Sodium polytungstate solution, ~85% in H2O, for separation of minerals
Sigma-Aldrich
Sodium polytungstate, purum p.a., for the preparation of heavy liquid, for sink-float analysis, ≥85% WO3 basis
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)