Skip to Content
Merck
All Photos(1)

Key Documents

71913

Sigma-Aldrich

Sodium polytungstate

purum p.a., for the preparation of heavy liquid, ≥85% WO3 basis

Synonym(s):

Sodium metatungstate, hexa-Sodium tungstate (W12)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3Na2WO4 · 9WO3 · H2O
CAS Number:
Molecular Weight:
2986.01
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

grade

purum p.a.

Assay

≥85% WO3 basis

form

powder or crystals

quality

for the preparation of heavy liquid

technique(s)

titration: suitable

color

white to off-white

pH

3 (20 °C)

density

3.1 g/cm3 at 20 °C (saturated aqueous solution)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O

InChI

1S/6Na.39O.12W/q6*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;6*-1;;;;;;;;;;;;

InChI key

AZCSOJKJFMWYCX-UHFFFAOYSA-N

Application

Sodium polytungstate (SPT) is widely used to produce heavy liquid for sink /float analysis (gravity separation) and density gradient centrifugation due to its very high solubility in water.

Other Notes

A new medium for density-gradient centrifugation, the density can be adjusted to up to 3.1 g/mlcitationcitation
A new medium for density-gradient centrifugation, the density can be adjusted to up to 3.1 g/ml

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Rothe et al.
PloS one, 10(11), e0143737-e0143737 (2015-11-26)
An increasing number of studies constrain the importance of iron for the long-term retention of phosphorus (P) under anoxic conditions, i.e. the formation of reduced iron phosphate minerals such as vivianite (Fe3(PO4)2⋅8H2O). Much remains unknown about vivianite formation, the factors
The use of low-toxic heavy suspensions in mineral sands evaluation and zircon fractionation.
Koroznikova L, et al.
Journal of the South African Veterinary Association, 25-33 (2008)
B. Plewinsky et al.
Makromol. Chem., 185, 1429-1429 (1984)
M.R. Gregory et al.
N.Z. J. Geol. Geophys., 30, 317-317 (1987)
Gregory M Hayes et al.
American journal of translational research, 7(6), 1181-1188 (2015-08-19)
Soft tissue sarcoma (STS) is a heterogenous tumor arising from the embryonic mesoderm represented by approximately 50 histological subtypes. Effective therapeutic intervention is lacking for recurrent, late stage and metastatic disease. CD39, a cell-surface ectonucleotidase, has previously been shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service