Merck
  • Nature of Li2O2 oxidation in a Li-O2 battery revealed by operando X-ray diffraction.
Nature of Li2O2 oxidation in a Li-O2 battery revealed by operando X-ray diffraction.
Journal of the American Chemical Society (2014-10-24)
Swapna Ganapathy, Brian D Adams, Georgiana Stenou, Maria S Anastasaki, Kees Goubitz, Xue-Fei Miao, Linda F Nazar, Marnix Wagemaker
ABSTRACT
Fundamental research into the Li-O2 battery system has gone into high gear, gaining momentum because of its very high theoretical specific energy. Much progress has been made toward understanding the discharge mechanism, but the mechanism of the oxygen evolution reaction (OER) on charge (i.e., oxidation) remains less understood. Here, using operando X-ray diffraction, we show that oxidation of electrochemically generated Li2O2 occurs in two stages, but in one step for bulk crystalline (commercial) Li2O2, revealing a fundamental difference in the OER process depending on the nature of the peroxide. For electrochemically generated Li2O2, oxidation proceeds first through a noncrystalline lithium peroxide component, followed at higher potential by the crystalline peroxide via a Li deficient solid solution (Li(2-x)O2) phase. Anisotropic broadening of the X-ray Li2O2 reflections confirms a platelet crystallite shape. On the basis of the evolution of the broadening during charge, we speculate that the toroid particles are deconstructed one platelet at a time, starting with the smallest sizes that expose more peroxide surface. In the case of in situ charged bulk crystalline Li2O2, the Li vacancies preferentially form on the interlayer position (Li1), which is supported by first-principle calculations and consistent with their lower energy compared to those located next to oxygen (Li2). The small actively oxidizing fraction results in a gradual reduction of the Li2O2 crystallites. The fundamental insight gained in the OER charge mechanism and its relation to the nature of the Li2O2 particles is essential for the design of future electrodes with lower overpotentials, one of the key challenges for high performance Li-air batteries.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Pricing and availability is not currently available.
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Pricing and availability is not currently available.
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Pricing and availability is not currently available.
Supelco
2-Propanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Pricing and availability is not currently available.