Merck
  • Chemical hazards present in liquids and vapors of electronic cigarettes.
Chemical hazards present in liquids and vapors of electronic cigarettes.
Archives of toxicology (2014-06-25)
Christoph Hutzler, Meike Paschke, Svetlana Kruschinski, Frank Henkler, Jürgen Hahn, Andreas Luch
ABSTRACT
Electronic (e-)cigarettes have emerged in recent years as putative alternative to conventional tobacco cigarettes. These products do not contain typical carcinogens that are present in tobacco smoke, due to the lack of combustion. However, besides nicotine, hazards can also arise from other constituents of liquids, such as solvents, flavors, additives and contaminants. In this study, we have analyzed 28 liquids of seven manufacturers purchased in Germany. We confirm the presence of a wide range of flavors to enhance palatability. Although glycerol and propylene glycol were detected in all samples, these solvents had been replaced by ethylene glycol as dominant compound in five products. Ethylene glycol is associated with markedly enhanced toxicological hazards when compared to conventionally used glycerol and propylene glycol. Additional additives, such as coumarin and acetamide, that raise concerns for human health were detected in certain samples. Ten out of 28 products had been declared "free-of-nicotine" by the manufacturer. Among these ten, seven liquids were identified containing nicotine in the range of 0.1-15 µg/ml. This suggests that "carry over" of ingredients may occur during the production of cartridges. We have further analyzed the formation of carbonylic compounds in one widely distributed nicotine-free brand. Significant amounts of formaldehyde, acetaldehyde and propionaldehyde were only found at 150 °C by headspace GC-MS analysis. In addition, an enhanced formation of aldehydes was found in defined puff fractions, using an adopted machine smoking protocol. However, this effect was delayed and only observed during the last third of the smoking procedure. In the emissions of these fractions, which represent up to 40 % of total vapor volume, similar levels of formaldehyde were detected when compared to conventional tobacco cigarettes. By contrast, carbonylic compounds were hardly detectable in earlier collected fractions. Our data demonstrate the necessity of standardized machine smoking protocols to reliably address putative risks of e-cigarettes for consumers.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
4-Bromofluorobenzene, analytical standard
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
1-Bromo-4-fluorobenzene, 99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O