Skip to Content
Merck
All Photos(1)

Key Documents

719099

Sigma-Aldrich

Acetaldehyde solution

5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021102
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

concentration

5 M in THF

refractive index

n20/D 1.389

density

0.868 g/mL at 20 °C
0.843 g/mL at 25 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(C)=O

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-25.1 °F - closed cup

Flash Point(C)

-31.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nikoline Borgermann et al.
The EMBO journal, 38(8) (2019-03-28)
DNA-protein crosslinks (DPCs) are highly cytotoxic lesions that obstruct essential DNA transactions and whose resolution is critical for cell and organismal fitness. However, the mechanisms by which cells respond to and overcome DPCs remain incompletely understood. Recent studies unveiled a
Marjie L Hard et al.
Placenta, 24(2-3), 149-154 (2003-02-05)
Significant interindividual variability exists following maternal alcohol consumption; not all children born to alcoholic women manifest the symptoms associated with foetal alcohol spectrum disorder (FASD). To investigate the potential role of the placenta as a source of variability by determining
Neil R McGregor
Alcohol (Fayetteville, N.Y.), 41(7), 469-478 (2007-11-06)
Recent introduction of several commercial Kudzu root (Pueraria lobata) containing hangover remedies has occurred in western countries. The available data is reviewed to assess if there are any potential concerns in relationship to the development of neoplasm if these products
Keitaro Matsuo et al.
Carcinogenesis, 34(7), 1510-1515 (2013-03-05)
The impact of alcohol on the risk of stomach cancer is controversial. Although aldehyde dehydrogenase 2 (ALDH2) Glu504Lys (rs671) polymorphism has a strong effect on acetaldehyde metabolism, little is known about its impact on stomach cancer risk when combined with
Dirk W Lachenmeier et al.
Addiction (Abingdon, England), 104(4), 533-550 (2009-04-02)
In addition to being produced in ethanol metabolism, acetaldehyde occurs naturally in alcoholic beverages. Limited epidemiological evidence points to acetaldehyde as an independent risk factor for cancer during alcohol consumption, in addition to the effects of ethanol. This study aims

Questions

  1. 購入を検討しています。 719099 Sigma-Aldrich アセトアルデヒド 溶液 50ml 上記の試薬はアセトアルデヒド原液でしょうか?

    1 answer
    1. This product is a 5 M Acetaldehyde solution in Tetrahydrofuran (THF) and may be considered a stock solution for some applications. Additional Acetaldehyde solutions are available in ethanol, water, and isopropranol. Please see the link below to review the available options:
      https://www.sigmaaldrich.com/substance/acetaldehydesolution440575070

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service