Merck
  • A comparison of two dental implant systems in partially edentulous patients: 4-month post-loading results from a pragmatic multicentre randomised controlled trial.
A comparison of two dental implant systems in partially edentulous patients: 4-month post-loading results from a pragmatic multicentre randomised controlled trial.
European journal of oral implantology (2013-08-09)
Marco Esposito, Rodolfo Blasone, Gianpaolo Favaretto, Claudio Stacchi, Matteo Calvo, Carlo Marin, Pietro Felice
ABSTRACT
To compare the clinical effectiveness of two implant systems: Way Milano and Kentron (Geass, Pozzuolo del Friuli, UD, Italy). A total of 64 patients requiring at least two single crowns or partial fixed dental prostheses supported by a maximum of three implants had their sites randomised according to a split-mouth design to receive both implant systems at six centres. Patients were followed up for 4 months after initial loading. Outcome measures were prosthesis/implant failures, any complication and clinician preference. In total, 71 Way Milano and 73 Kentron implants were placed. One patient died 45 days after placement of 2 implants, but all remaining patients were followed up to 4 months post-loading. No Way Milano implant failed whereas 3 Kentron implants failed before loading. Two complications were reported, one at each implant type. There were no statistically significant differences for prosthesis/ implant success and complications between the implant systems. Three operators preferred Way Milano implants whereas the other 3 had no preference. Preliminary short-term data (4 months post-loading) showed no statistically significant differences between the two implant systems, however trends were suggestive of a better clinical performance for Way Milano implants.
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance