Skip to Content
Merck
All Photos(1)

Key Documents

C8919

Sigma-Aldrich

Bovine Collagen Type I

Collagen from calf skin

from bovine skin, liquid, 1 mg/mL, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture

biological source

bovine (calf) skin

Quality Level

sterility

aseptically processed

product line

BioReagent

form

solution (0.1% solution in 0.1 M acetic acid)

packaging

pkg of 20 mL

concentration

(0.1% solution in 0.1 M acetic acid)

technique(s)

cell culture | mammalian: suitable

surface coverage

6‑10 μg/cm2

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

Peptide Source: Elastin

Peptide Source: Fibronectin

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Collagen type I is a component of skin, bone, tendon, and other fibrous connective tissues. It consists of repeating triplet amino acids glycine, proline and hydroxyproline. Collagen is a left handed helix with three polypeptide chains.[1] It is a component of extracellular matrix and as close to 28 types are present in bovine.[2]

Application

Collagen from calf skin has been used:
  • for pre-coting glass slides for immunofluorescence studies[3]
  • as a cell adhesion factor and modification of poly(vinylidene fluoride-trifluoroethylene) (P(VDF-TrFE)) films for neuron culture[4]
  • for coating culture dishes for murine embryonic fibroblasts culture[5]

This product is intended to produce thin layer coatings on tissue culture plates to facilitate attachment of anchorage-dependent cells, recommended for use at 6-10 μg/cm2. It is NOT intended for production of 3-D gels. Type I collagen is often used in cell culture as an attachment substratum with myoblasts, spinal ganglia, hepatocytes, embryonic lung, heart explants, fibroblasts, endothelial cells, and islet cells have all been cultured successfully on films or gels of type I collagen. Collagen type I may also be used in research of Idiopathic pulmonary fibrosis (IPF), studies on the effect of ER stress IPF on lung fibroblasts. Collagen in acidic solution can produce three dimensional scaffolding with use in bioengineering and cell culture applications.

Biochem/physiol Actions

Collagen type I on heat denaturation results in disruption of triple helix to a randomly coil.[1] Mutations in the collagen gene are implicated in a variety of cattle diseases.[2] It has applications in food and cosmetics. Collagen is used as a biomaterial and as a tissue scaffold in tissue engineering.[6] Collagen type I fiber assembly is very crucial for physiological processes and cellular signaling.[7]

Components

All collagen molecules are composed of three polypeptide chains arranged in a triple helical conformation, with a primary structure that is mostly a repeating motif with glycine in every third position and proline or 4-hydroxyproline frequently preceding the glycine residue. Type I collagen differs from other collagens by its low lysine hydroxylation and low carbohydrate composition.

Preparation Note

This product is a 0.1% (1mg/ml) solution of calf skin collagen 0.1 M acetic acid. The product′s appearance is turbid, milky white solution. It is recommended that dilutions of product are prepared in sterile water. Please refer to information given on the product information sheet prior to introduction of product to cells and media.

Other Notes

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extracellular Collagen VI Has Prosurvival and Autophagy Instructive Properties in Mouse Fibroblasts
Castagnaro S, et al.
Frontiers in Physiology, 9(3), 1129-1129 (2018)
Vahid Serpooshan et al.
Scientific reports, 7(1), 12590-12590 (2017-10-05)
During normal lifespan, the mammalian heart undergoes limited renewal of cardiomyocytes. While the exact mechanism for this renewal remains unclear, two possibilities have been proposed: differentiated myocyte replication and progenitor/immature cell differentiation. This study aimed to characterize a population of
Collagen: a network for regenerative medicine
Pawelec KM, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 4(40), 6484-6496 (2016)
Hedgehog antagonist cyclopamine isomerizes to less potent forms when acidified
Wilson SR, et al.
Journal of Pharmaceutical and Biomedical Analysis, 52(5), 707-713 (2010)
Culturing neurons on MEMS fabricated P (VDF-TrFE) films for implantable artificial cochlea
Shintaku H, et al.
Journal of Biomechanical Science and Engineering, 5(3), 229-235 (2010)

Articles

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

See All

Questions

1–8 of 8 Questions  
  1. i want to prepare nanofibers from collagen, then what type of collagen I want to use.

    1 answer
    1. This product has not been tested for use in creating collagen nanofibers. The literature suggests that a Bornstein and Traub Type I Collagen would be suitable. As the product is a dilute solution at acidic pH, a powdered material may be a preferred option. The end user would have to determine suitability. Please see the link below to a publication that successfully used product C9791 in the creation of collagen nanofibers:
      https://www.sciencedirect.com/science/article/pii/S0378517317307780

      Please see the link below to review this product, which is the starting material for C8919:
      https://www.sigmaaldrich.com/product/sigma/c9791

      Helpful?

  2. Product C8919, Collagen solution, gelled while refrigerated.  Can it still be used?

    1 answer
    1. This can occur but does not harm the product.  When warmed to room temperature, the product will liquify.  Please make sure it is well mixed prior to dilution for coating cultureware.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How can I sterilize collagen solutions?

    1 answer
    1. We recommend transferring the collagen solution to a glass bottle with a screw cap and carefully layering chloroform at the bottom. The amount of chloroform to use should be ~10% of the volume of collagen solution. DO NOT SHAKE OR STIR. Allow to stand overnight at 2-8 °C. Aseptically remove the top layer containing the collagen solution to a new bottle. We do not recommend sterilizing the collagen solution by membrane filtration. We have found substantial protein loss by this method.

      Helpful?

  5. Can Product C8919, Collagen solution, be used to make 3-D collagen gels?

    1 answer
    1. This product is NOT intended to produce 3-D (thick) gels, since the solution will not gel. This may be due to gamma-irradiation of the powder during the production process.  We recommend using Product No C4243, which is tested and found to be suitable for 3-D gel formation.

      Helpful?

  6. How can I use Product C8919, Collagen solution, to coat my cultureware?

    1 answer
    1. A protocol for coating cultureware can be found on the product information sheet (under Documents, above).

      Helpful?

  7. How long can plates coated with Product C8919, Collagen solution, be stored?

    1 answer
    1. We make no recommendations about storage of plates after coating.  Since factors such as humidity may impact on longevity of coated plates, customers should make their own determination as to whether plates can be stored and under which conditions.  We used to offer SigmaScreen collagen coated plates as Product No. S3315. For optimal performance of that product, we recommend that the unopened product should be stored in a dry place at 2-8°C. We found that the S3315 could be stored at room temperature for up to three months, and that it should never be exposed to temperatures above 50°C.

      Helpful?

  8. Why does Product C8919, Collagen solution, look milky or contain particulates?

    1 answer
    1. C8919, Collagen Solution from calf skin, is prepared by allowing irradiated sterile collagen strands to mix with sterile filtered 0.1N acetic acid over several hours. The collagen dissolves to an opalescent solution that is aseptically dispensed while stirring.  No further filtration is necessary.  Due to cross-linkage and sequence variability, as well as structural changes that can occur during isolation and purification, Collagen Type I (Bornstein and Traub) is a heterogeneous mixture rather than a pure compound.  This variability, along with slight differences in the raw material, lends itself to shifts in the opacity of the solution from lot to lot.  This product is tested for sterility prior to release for sale to our customers.  A milky or particulate appearance is not indicative of microbial contamination and does not affect product use. .

      Helpful?

Reviews

Active Filters

  1. Campinas - Brazil
    • Reviews 2
    • Votes 0
    3 out of 5 stars.

    Of its use for platelet aggregation

    Collagens are often used to assess platelet aggregation. This collagen did not induce a platelet response in concentrations as high as 100 ug/mL and therefore is not suitable to study platelet function. This can be used to assess collagen fibrillogenesis, although the curve is much more evident when using rat tail collagen.

    Helpful?

    1. Response from MilliporeSigma:

      C8919 is a collagen solution at 0.03 - 0.15 % in 0.1M acetic acid and is not pure collagen. Review your lot specific COA in the Documentation section above for the % value of your lot. Please review your calculations to determine if the solution was diluted properly.
      We encourage customers who experience problems with our products to contact us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service