Skip to Content
Merck
All Photos(4)

Key Documents

C9879

Sigma-Aldrich

Bovine Collagen Type I

from bovine achilles tendon, powder, suitable for substrate for collagenase

Synonym(s):

Collagen powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

Product Name

Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase

biological source

bovine Achilles tendon

form

powder

technique(s)

ELISA: suitable
activity assay: suitable

suitability

suitable for substrate for collagenase

UniProt accession no.

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Collagen is a widely expressed protein in the body. There are 20 different types of collagen in human tissue. Type I collagen is the abundant bone protein. It constitutes ~90% of bone organic matter.
Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.
Collagen terminology using the Bornstein and Traub designation originates from the reference; Bornstein, P. and Traub, W. The Proteins, (1979) 4, 411-605

Application

Bovine achilles tendon collagen was used in a study of plant extracts as inhibitors of MMP-collagenases, the putative active agents in the breakdown of cartilage in osteoarthritis and rheumatoid arthritis.
Collagen from bovine achilles tendon is suitable for use in:
  • the detection of collagenase activity
  • as a reference sample in the thermal analysis study of human bone using differential scanning calorimetry, thermogravimetry, gas chromatography and Fourier transform infrared spectroscopy
  • as a substrate for developing a simple assay for determining collagen degradation in vitro
  • a study to examine the binding activity of the integral glycoprotein dipeptidyl peptidase IV to insoluble type I collagen by solid-phase enzyme-linked immunosorbent assay
Collagen from bovine Achilles tendon is a naturally occurring protein in the form of elongated fibrils. It may be used in studies of the fibrocartilaginous zone, which the collagen must first pass through before inserting into the calcaneus. It may also be used in studies of growth factor effects on collagen content and cross-linking during Achilles tendon healing.

Biochem/physiol Actions

Collagen from bovine Achilles tendon is a naturally occurring protein in the form of elongated fibrils. It may be used in studies of the fibrocartilaginous zone, which the collagen must first pass through before inserting into the calcaneus. It may also be used in studies of growth factor effects on collagen content and cross-linking during Achilles tendon healing.
Collagen is an insoluble fibrous protein that is part of the extracellular matrix and the connective tissue. It is responsible for the ability of the tissues to withstand stretching. The long precursors called procollagens are synthesized and assembled in the ER, secreted into the extracellular space and processed to form collagen fibres. The different types of collagen are composed of molecules containing three polypeptide chains arranged in a triple helical conformation varying slightly in the amino acid sequence. The primary structure is a repeating motif with glycine in every third position preceded frequently with a proline or 4-hydroxyproline residue.

Preparation Note

Prepared by the method of Einbinder, J. and Schubert, M., J. Biol. Chem., 188, 335 (1951).

Reconstitution

This product is an insoluble collagen preparation. It is insoluble in water, aqueous buffers, dilute acid, and organic solvents. For use as a substrate in collagenase assays, this collagen can be prepared as a suspension in 50 mM TES buffer, pH 7.4 with 0.36 mM calcium chloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E K Steffen et al.
Journal of clinical microbiology, 14(2), 153-156 (1981-08-01)
Thirty-three strains of anaerobic bacteria isolated from human clinical specimens were examined for the presence of heparinase, hyaluronidase, chondroitin sulfatase, gelatinase, collagenase, fibrinolysin, lecithinase, and lipase activities. Pronounced heparinase activity was limited to species of the genus Bacteroides. A number
Extraction and partial characterization of collagen from different animal skins
Quereshi S, et al.
Recent Research in Science and Technology, 2(9) (2010)
Thermal analysis study of human bone
Lozano L F, et al.
J. Mater. Sci., 38, 4777-4782 (2003)
M L Tanzer
Science (New York, N.Y.), 180(4086), 561-566 (1973-05-11)
The formation of collagen cross-links is attributable to the presence of two aldehyde-containing amino acids which react with other amino acids in collagen to generate difunctional, trifunctional, and tetrafunctional cross-links. A necessary prerequisite for the development of these cross-links is
Bradley E Layton et al.
Journal of molecular evolution, 66(6), 539-554 (2008-06-04)
Two competing effects at two vastly different scales may explain collagen's current translation length. The necessity to have long molecules for maintaining mechanical integrity at the organism and supraorganism scales may be limited by the need to have small molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service