Merck
[Novel autoantibodies in myasthenia gravis].
Nihon rinsho. Japanese journal of clinical medicine (2013-06-20)
Akiko Nagaishi, Waka Sakai, Masakatsu Motomura
ABSTRACT
Patients with myasthenia gravis(MG) are divided into three groups: (1) acetylcholine receptor antibody positive MG: 80%, (2) muscle-specific receptor tyrosine kinase (MuSK) antibody positive MG: 5-10%, and (3) double seronegative MG. In 2011, autoantibodies (Abs) against low-density lipoprotein receptor-related protein 4(Lrp4) were identified in Japanese MG patients and thereafter have been reported in Germany and USA. In other Lrp4 Ab papers, Lrp4 Ab positive sera inhibited agrin-induced aggregation of AChRs in cultured myotubes, suggesting a pathogenic role regarding the dysfunction of the neuromuscular endplate. Anti-MuSK autoantibodies were revealed to block binding of collagen Q (ColQ) to MuSK. Anti-Kv1.4 antibodies targeting alpha-subunits(Kv1.4) of the voltage-gated potassium channel occurs frequently among MG patients with thymoma. Further understandings of neuromuscular junction structure and functions through newly discovered autoantibodies may provide more specific clinical information and treatments in MG.
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture