Merck
  • Chloroplast movement provides photoprotection to plants by redistributing PSII damage within leaves.
Chloroplast movement provides photoprotection to plants by redistributing PSII damage within leaves.
Photosynthesis research (2012-06-15)
Phillip A Davis, Roger P Hangarter
ABSTRACT
Plants use light to fix carbon through the process of photosynthesis but light also causes photoinhibition, by damaging photosystem II (PSII). Plants can usually adjust their rate of PSII repair to equal the rate of damage, but under stress conditions or supersaturating light-intensities damage may exceed the rate of repair. Light-induced chloroplast movements are one of the many mechanisms plants have evolved to minimize photoinhibition. We found that chloroplast movements achieve a measure of photoprotection to PSII by altering the distribution of photoinhibition through depth in leaves. When chloroplasts are in the low-light accumulation arrangement a greater proportion of PSII damage occurs near the illuminated surface than for leaves where the chloroplasts are in the high-light avoidance arrangement. According to our findings chloroplast movements can increase the overall efficiency of leaf photosynthesis in at least two ways. The movements alter light profiles within leaves to maximize photosynthetic output and at the same time redistribute PSII damage throughout the leaf to reduce the amount of inhibition received by individual chloroplasts and prevent a decrease in photosynthetic potential.
Product Description
Lincomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lincomycin hydrochloride monohydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Lincomycin hydrochloride, 96.0-102.0% (HPLC)
Sigma-Aldrich
Lincomycin hydrochloride, ≥90% (TLC)
Sigma-Aldrich
Lincomycin hydrochloride, BioReagent, suitable for cell culture