Skip to Content
Merck
All Photos(1)

Key Documents

L0650000

Lincomycin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lincomycin hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34N2O6S · HCl · H2O
CAS Number:
Molecular Weight:
461.01
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lincomycin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].[H]O[H].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H34N2O6S.ClH.H2O/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H;1H2/t9-,10-,11+,12-,13+,14-,15-,16-,18-;;/m1../s1

InChI key

LFZGYTBWUHCAKF-DCNJEFSFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lincomycin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Chih Tseng et al.
Plant molecular biology, 82(4-5), 375-392 (2013-05-07)
RNA editing is one of the post-transcriptional processes that commonly occur in plant plastids and mitochondria. In Arabidopsis, 34 C-to-U RNA editing events, affecting transcripts of 18 plastid genes, have been identified. Here, we examined the editing and expression of
Ulrich Schreiber et al.
Photosynthesis research, 114(3), 165-177 (2013-02-15)
A new type of multi-color PAM chlorophyll fluorometer (Schreiber et al. 2012) was applied for measurements of photodamage to photosystem II (PS II) in optically thin suspensions of Chlorella (200 μg Chl l(-1)) in the presence of 1 mM lincomycin.
Kazunori Miyata et al.
Photosynthesis research, 113(1-3), 165-180 (2012-07-17)
When visible light is excess, the photosynthetic machinery is photoinhibited. The extent of net photoinhibition of photosystem II (PSII) is determined by a balance between the rate of photodamage to D1 and some other PSII proteins and the rate of
C F Williams et al.
Journal of environmental quality, 41(5), 1473-1480 (2012-10-27)
The reuse of treated wastewater for groundwater recharge is an effective way to provide advanced treatment and water storage. Contaminants such as human drugs have been identified as a potential problem for use of this water. Gilbert, Arizona maintains a
Phillip A Davis et al.
Photosynthesis research, 112(3), 153-161 (2012-06-15)
Plants use light to fix carbon through the process of photosynthesis but light also causes photoinhibition, by damaging photosystem II (PSII). Plants can usually adjust their rate of PSII repair to equal the rate of damage, but under stress conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service