Skip to Content
Merck
All Photos(3)

Key Documents

L2774

Sigma-Aldrich

Lincomycin hydrochloride

suitable for cell culture, BioReagent

Synonym(s):

Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-erythro-α-D-galactooctopyranoside hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34N2O6S · HCl
CAS Number:
Molecular Weight:
443.00
Beilstein:
4171650
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

product name

Lincomycin hydrochloride, BioReagent, suitable for cell culture

product line

BioReagent

Quality Level

potency

800-900 units per mg

technique(s)

cell culture | mammalian: suitable

color

white to yellow-white

solubility

H2O: 4 mL, clear, colorless to faintly yellow (200 mg + 4 mL H2O)

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H34N2O6S.ClH/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H/t9-,10-,11+,12-,13+,14-,15-,16-,18-;/m1./s1

InChI key

POUMFISTNHIPTI-BOMBIWCESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Lincomycin hydrochloride is an antibiotic used to study the inhibition of protein synthesis, bacterial antibiotic resistance and protein and surfactant binding. It is produced by Streptomyces lincolnensis var. lincolnensis, and has been used in the treatment of Staphylococcal, Streptococcal, and Bacterioides fragilis infections.

Biochem/physiol Actions

Mode of action: Lincomycin inhibits bacterial protein synthesis by forming cross-links within the peptidyl transferase loop region of the 23S rRNA.†

Antimicrobial spectrum: Lincomycin hydrochloride is effective against gram-positive bacteria.

Preparation Note

This product is soluble in water at 50 mg/mL, yielding a clear, colorless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu Duan et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 375(1801), 20190402-20190402 (2020-05-05)
Chloroplast-to-nucleus retrograde signalling (RS) is known to impact plant growth and development. In Arabidopsis, we and others have shown that RS affects seedling establishment by inhibiting deetiolation. In the presence of lincomycin, a chloroplast protein synthesis inhibitor that triggers RS
Yongjian Qiu et al.
Nature communications, 10(1), 140-140 (2019-01-13)
Ambient temperature sensing by phytochrome B (PHYB) in Arabidopsis is thought to operate mainly at night. Here we show that PHYB plays an equally critical role in temperature sensing during the daytime. In daytime thermosensing, PHYB signals primarily through the
Phillip A Davis et al.
Photosynthesis research, 112(3), 153-161 (2012-06-15)
Plants use light to fix carbon through the process of photosynthesis but light also causes photoinhibition, by damaging photosystem II (PSII). Plants can usually adjust their rate of PSII repair to equal the rate of damage, but under stress conditions
Kazunori Miyata et al.
Photosynthesis research, 113(1-3), 165-180 (2012-07-17)
When visible light is excess, the photosynthetic machinery is photoinhibited. The extent of net photoinhibition of photosystem II (PSII) is determined by a balance between the rate of photodamage to D1 and some other PSII proteins and the rate of
C F Williams et al.
Journal of environmental quality, 41(5), 1473-1480 (2012-10-27)
The reuse of treated wastewater for groundwater recharge is an effective way to provide advanced treatment and water storage. Contaminants such as human drugs have been identified as a potential problem for use of this water. Gilbert, Arizona maintains a

Articles

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service