Merck
  • [Active neuraminidase constituents of Polygonum cuspidatum against influenza A(H1N1) influenza virus].
[Active neuraminidase constituents of Polygonum cuspidatum against influenza A(H1N1) influenza virus].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-01-15)
Kao-Tan Chen, Wei-Ling Zhou, Jia-Wei Liu, Mian Zu, Zi-Ning He, Guan-Hua Du, Wei-Wen Chen, Ai-Lin Liu
ABSTRACT
To isolate and identify active neuraminidase constituents of Polygonum cuspidatum against influenza A (H1N1) influenza virus. On the basis of the bioassay-guided fractionation,such chromatographic methods as silica gel, sephadex LH-20 and HPLC were adopted to isolate active constituents of extracts from Polygonum cuspidatum, and their molecular structures were identifiied on the basis of their spectral data such as NMR and MS and physico-chemical properties. Seven compounds were isolated from the ethyl acetate extract of P. cuspidatum and identified as 2-methoxystypandrone (1), emodin (2), resveratrol (3), polydatin (4), emodin-8-O-beta-D-glucopyranoside (5), (E)-3, 5, 12-trihydroxystilbene-3-O-beta-D-glucopyranoside-2'-(3", 4", 5"-trihydroxybenzoate) (6) and catechin-3-O-gallate (7), respectively. Among them, the NA test showed that compounds 3, 6 and 7 had inhibitory effect against NAs activity, with IC50 values of 129.8, 44.8 and 21.3 micromol x L(-1), respectively. Moreover, the further CPE test showed compounds 6 and 7 had significant inhibitory effect against H1N influenza virus (EC50 = 5.9, 0.9 micromol x L(-1), respectively), with very low cytotoxicity to the host cells, their therapeutic selective index(SI) in MDCK cells ranged from 56 to 269. The neuraminidase inhibitors against H1N1 anti-influenza virus isolated from extracts of P. cuspidatum on the basis of the bioassay-guided fractionation are significant in specifying their therapeutic material basis and drug R&D against influenza.
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)