Merck
  • Chemerin regulates normal angiogenesis and hypoxia-driven neovascularization.
Chemerin regulates normal angiogenesis and hypoxia-driven neovascularization.
Angiogenesis (2021-09-16)
Cyrine Ben Dhaou, Kamel Mandi, Mickaël Frye, Angela Acheampong, Ayoub Radi, Benjamin De Becker, Mathieu Antoine, Nicolas Baeyens, Valérie Wittamer, Marc Parmentier
ABSTRACT
Chemerin is a multifunctional protein initially characterized in our laboratory as a chemoattractant factor for leukocyte populations. Its main functional receptor is CMKLR1. We identified previously chemerin as an anti-tumoral factor inhibiting the vascularization of tumor grafts. We show here that overexpression of bioactive chemerin in mice results in a reduction of the density of the retinal vascular network during its development and in adults. Chemerin did not affect vascular sprouting during the post-natal development of the network, but rather promoted endothelial cell apoptosis and vessel pruning. This phenotype was reversed to normal in CMKLR1-deficient mice, demonstrating the role of this receptor. Chemerin inhibited also neoangiogenesis in a model of pathological proliferative retinopathy, and in response to hind-limb ischemia. Mechanistically, PTEN and FOXO1 antagonists could almost completely restore the density of the retinal vasculature, suggesting the involvement of the PI3-kinase/AKT pathway in the chemerin-induced vessel regression process.
Sigma-Aldrich
bpV(HOpic), A potent phosphotyrosine phosphatase inhibitor.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Millipore
FluorSave Reagent
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Foxo1 Inhibitor, AS1842856, Foxo1 Inhibitor, AS1842856, is a cell-permeable inhibitor that blocks the transcription activity of Foxo1 (IC₅₀ = 33 nM). Directly binds to the active Foxo1, but not the Ser256-phosphorylated form.