Skip to Content
Merck
All Photos(1)

Key Documents

108014

Sigma-Aldrich

Benzyl ether

98%

Synonym(s):

Dibenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
Beilstein:
1911156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

refractive index

n20/D 1.561 (lit.)

bp

298 °C (lit.)

mp

1.5-3.5 °C (lit.)

solubility

H2O: insoluble
acetone: miscible
chloroform: miscible
diethyl ether: miscible
ethanol: miscible

density

1.043 g/mL at 25 °C (lit.)

functional group

ether
phenyl

SMILES string

C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H14O/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

MHDVGSVTJDSBDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl ether is an aromatic ether that is commonly employed as a protecting agent and benzylating agent.[1][2]

Benzyl ether is a plasticizer for nitrocellulose and a solvent in perfumery[3].

Application

Benzyl ether was used as an effective GFP friendly tissue clearing medium for mouse brains[4]. It was used in a study to develop a detailed protocol for performing 3D imaging of solvent-cleared organs and its application to various microscopy techniques[5].

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot synthesis of mesoporous ZrPW solid acid catalyst for liquid phase benzylation of anisole
Miao Z, et al.
Catalysis Science & Technology, 4, 838-850 (2014)
Evaluation of the Genetic Toxicity of Synthetic Chemicals (XIV)-in vitro Chromosomal Aberration Assay with 11 Chemicals in Chinese Hamster Lung Cells.
Kim YJ and Ryu JC.
Molecular & Cellular Toxicology, 2(2), 89-96 (2006)
Active cobalt catalyst for the cleavage of benzyl ether
Wang B, et al.
The Journal of Organic Chemistry, 76, 9531-9535 (2011)
Klaus Becker et al.
PloS one, 7(3), e33916-e33916 (2012-04-06)
Generally, chemical tissue clearing is performed by a solution consisting of two parts benzyl benzoate and one part benzyl alcohol. However, prolonged exposure to this mixture markedly reduces the fluorescence of GFP expressing specimens, so that one has to compromise
Ya-Ping Tu
The Journal of organic chemistry, 71(15), 5482-5488 (2006-07-15)
It is often found in mass spectrometry that when a molecule is protonated at the thermodynamically most favorable site, no fragmentation occurs, but a major reaction is observed when the proton migrates to a different position. For benzophenones, acetophenones, and

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service