Merck
  • Discovery of anti-inflammatory physiological peptides that promote tissue repair by reinforcing epithelial barrier formation.
Discovery of anti-inflammatory physiological peptides that promote tissue repair by reinforcing epithelial barrier formation.
Science advances (2021-11-18)
Yukako Oda, Chisato Takahashi, Shota Harada, Shun Nakamura, Daxiao Sun, Kazumi Kiso, Yuko Urata, Hitoshi Miyachi, Yoshinori Fujiyoshi, Alf Honigmann, Seiichi Uchida, Yasushi Ishihama, Fumiko Toyoshima
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Anti-α-Catenin antibody produced in rabbit, whole antiserum
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α1-Antitrypsin from human plasma, salt-free, lyophilized powder