Merck
  • Synthesis of glycopeptide dendrimers, dimerization and affinity for Concanavalin A.
Synthesis of glycopeptide dendrimers, dimerization and affinity for Concanavalin A.
Bioorganic & medicinal chemistry (2011-04-13)
Ronan Euzen, Jean-Louis Reymond
ABSTRACT
We described herein the synthesis of second generation glycopeptide dendrimers G2a-g presenting variable amino acids placed internally into the multivalent scaffold. The effect of such structural modulation on recognition processes by Concanavalin A (Con A), was then estimated by enhanced-sensitivity Enzyme-Linked Lectin Assay (ELLA). In a complementary study, glycopeptide dendrons of different valencies and including a l-cysteine residue before the dendritic core (G0SH, G1SH and G2SH), were also synthesized and homodimerized. Then, the disulfide-containing glycopeptide dendrimers generated by this convergent approach (G0(2)S(2), G1(2)S(2) and G2(2)S(2)) were used as Con A inhibitors and assayed by ELLA.
Product Description
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
4-Nitrophenyl α-D-mannopyranoside, α-mannosidase substrate
Sigma-Aldrich
Methyl α-D-mannopyranoside, ≥99.0% (HPLC)
Millipore
Methyl α-D-mannopyranoside, ≥99.0%, suitable for microbiology, enables differentiation between species of Listeria