Skip to Content
Merck

Skip To

63579

D-(+)-Mannose

BioUltra, ≥99.5% (sum of enantiomers, HPLC)

Synonym(s):

D-Mannopyranose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
222-392-4
Beilstein/REAXYS Number:
1564373
MDL number:
eCl@ss:
32160703
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

assay

≥99.5% (sum of enantiomers, HPLC)

form

solid

optical activity

[α]20/D +13.8±0.5°, 24 hr, c = 10% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

mp

133-140 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤200 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.5 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤200 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤20 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.1, λ: 280 nm Amax: 0.1

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

application(s)

advanced drug delivery

storage temp.

room temp

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-QTVWNMPRSA-N

Application

D-(+)-Mannose, a C-2 epimer of D-glucose, is used in the formation of glycan structure and glycosylation.

Biochem/physiol Actions

Mannose, a six-carbon carbohydrate, is the C-2 epimer of glucose and a critical sugar for protein glycosylation. Mannose can also be utilized by the brain as an alternative energy source.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M2069M857492683
description

BioUltra, ≥99.5% (sum of enantiomers, HPLC)

description

≥99% (GC), wood

description

synthetic, ≥99% (GC)

description

analytical standard

assay

≥99.5% (sum of enantiomers, HPLC)

assay

≥99% (GC)

assay

≥99% (GC)

assay

≥95.0% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

solubility

H2O: 1 M, clear, colorless

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

water: soluble 10 g/L at 20 °C

solubility

-

form

solid

form

powder

form

powder

form

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

-

mp

133-140 °C (lit.)

mp

133-140 °C (lit.)

mp

133-140 °C (lit.)

mp

133-140 °C (lit.)


Still not finding the right product?

Explore all of our products under D-(+)-Mannose


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters