Merck
  • Magnetically-orientable Tween-based model membranes for NMR studies of proteins.
Magnetically-orientable Tween-based model membranes for NMR studies of proteins.
Biochimica et biophysica acta. Biomembranes (2020-06-01)
Andrée E Gravel, Alexandre A Arnold, Matthieu Fillion, Michèle Auger, Dror E Warschawski, Isabelle Marcotte
ABSTRACT
We present a new membrane mimetic system using a membrane softening detergent commonly known as Tween 80 (TW80), to form oriented systems for solid-state NMR applications. TW80 is a fatty acid ester (oleate) of sorbitan polyethoxylate and a mild non-ionic surfactant. Phosphatidylcholine (PC)/TW80 model membrane systems were characterized by solid-state NMR and FTIR spectroscopy. 31P and 2H NMR spectra showed that DMPC (14:0) and DPPC (16:0) self-assemble with TW80 to form oriented structures, and maintain alignment over a wide range of molar ratios and temperatures. The addition of lanthanide ions revealed that the membrane alignment can be flipped from parallel to perpendicular with respect to the magnetic field direction. Using 15N solid-state NMR and a labeled model transmembrane peptide, we showed that TW80-based membranes can be employed to determine the peptide orientation in the magnetic field, which is useful for structural determination. Altogether, our work showed that TW80 could be exploited for direct and efficient membrane protein extraction and to enhance membrane and membrane protein orientation without using a detergent removal step. This approach could be extended to a wide range of membranes including native ones.
Product Description
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 PC-d62, Avanti Research - A Croda Brand 860355C
Avanti
14:0 PC-d54, 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, powder
Avanti
16:0 PC-d62, Avanti Research - A Croda Brand 860355P, powder
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0 PC-d54, 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder