Merck
  • Enhancement of human iPSC-derived cardiomyocyte maturation by chemical conditioning in a 3D environment.
Enhancement of human iPSC-derived cardiomyocyte maturation by chemical conditioning in a 3D environment.
Journal of molecular and cellular cardiology (2019-10-28)
Chen Yu Huang, Rebeca Peres Moreno Maia-Joca, Chin Siang Ong, Ijala Wilson, Deborah DiSilvestre, Gordon F Tomaselli, Daniel H Reich
ABSTRACT
Recent advances in the understanding and use of pluripotent stem cells have produced major changes in approaches to the diagnosis and treatment of human disease. An obstacle to the use of human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs) for regenerative medicine, disease modeling and drug discovery is their immature state relative to adult myocardium. We show the effects of a combination of biochemical factors, thyroid hormone, dexamethasone, and insulin-like growth factor-1 (TDI) on the maturation of hiPSC-CMs in 3D cardiac microtissues (CMTs) that recapitulate aspects of the native myocardium. Based on a comparison of the gene expression profiles and the structural, ultrastructural, and electrophysiological properties of hiPSC-CMs in monolayers and CMTs, and measurements of the mechanical and pharmacological properties of CMTs, we find that TDI treatment in a 3D tissue context yields a higher fidelity adult cardiac phenotype, including sarcoplasmic reticulum function and contractile properties consistent with promotion of the maturation of hiPSC derived cardiomyocytes.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dodecenylsuccinic anhydride, mixture of isomers, technical grade, 90%
Sigma-Aldrich
Methyl-5-norbornene-2,3-dicarboxylic anhydride, 90%
Sigma-Aldrich
Triton X-100, laboratory grade