443271
Dodecenylsuccinic anhydride, mixture of isomers
technical grade, 90%
Synonym(s):
DDSA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
technical grade
Quality Level
Assay
90%
form
liquid
refractive index
n20/D 1.479 (lit.)
bp
150 °C/3 mmHg (lit.)
density
1.005 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCC\C=C\C1CC(=O)OC1=O
InChI
1S/C16H26O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15(17)19-16(14)18/h11-12,14H,2-10,13H2,1H3/b12-11+
InChI key
WVRNUXJQQFPNMN-VAWYXSNFSA-N
General description
Dodecenylsuccinicanhydride (DDSA) is a mixture of isomers that belongs to the class of long-chained aliphatic anhydrides. It is composed of a dodecenyl (C12) chain attached to a succinic acid backbone and can undergo esterification, amidation, and ring-openingreactions. It is widely used as a cross-linking agent to synthesize hydrogels for drug delivery, coating for medical devices, and as a hardener for curing epoxy resins.
Application
Dodecenylsuccinic anhydride, mixture of isomers can be used:
- As a precursor to synthesize modified chitosan hydrogels useful for the sustained local delivery of drugs. This modification enhances the hydrophobicity of chitosan, allowing it to better encapsulate and retain hydrophobic drugs.
- To prepare high-performance Arabinoglucuronoxylan (AGX)-based biosurfactants using rapid homogeneous esterification reactions. These surfactants can be used for oily sludge remediation and oil recovery.
- As a hardener for curing thermosetting epoxy resins used in various fields such as paints, corrosion protective coatings, adhesives, and electrical circuits.
- As a modifying agent to prepare collagen hydrogels to fabricate skin wound dressings.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
High-performance arabinoglucuronoxylan-based biosurfactants for oily sludge separation
Haibo Huang, et al.
Carbohydrate Polymers, 303, 120461-120461 (2023)
Glycine surfactants derived from dodecenyl succinic anhydride
Dix L and Moon A
Journal of Surfactants and Detergents, 15(3), 351-357 (2012)
Bancroft's Theory and Practice of Histological Techniques (2018)
Epoxy resins
Nixon R, et al.
Kanerva's Occupational Dermatology, 559-581 (2012)
Magnetic microreactors for efficient and reliable magnetic nanoparticle surface functionalization
Digigow RG, et al.
Lab on a chip, 14(13), 2276-2286 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service