Merck
  • Torin2 Exploits Replication and Checkpoint Vulnerabilities to Cause Death of PI3K-Activated Triple-Negative Breast Cancer Cells.
Torin2 Exploits Replication and Checkpoint Vulnerabilities to Cause Death of PI3K-Activated Triple-Negative Breast Cancer Cells.
Cell systems (2019-12-10)
Sameer S Chopra, Anne Jenney, Adam Palmer, Mario Niepel, Mirra Chung, Caitlin Mills, Sindhu Carmen Sivakumaren, Qingsong Liu, Jia-Yun Chen, Clarence Yapp, John M Asara, Nathanael S Gray, Peter K Sorger
ABSTRACT
Frequent mutation of PI3K/AKT/mTOR signaling pathway genes in human cancers has stimulated large investments in targeted drugs but clinical successes are rare. As a result, many cancers with high PI3K pathway activity, such as triple-negative breast cancer (TNBC), are treated primarily with chemotherapy. By systematically analyzing responses of TNBC cells to a diverse collection of PI3K pathway inhibitors, we find that one drug, Torin2, is unusually effective because it inhibits both mTOR and other PI3K-like kinases (PIKKs). In contrast to mTOR-selective inhibitors, Torin2 exploits dependencies on several kinases for S-phase progression and cell-cycle checkpoints, thereby causing accumulation of single-stranded DNA and death by replication catastrophe or mitotic failure. Thus, Torin2 and its chemical analogs represent a mechanistically distinct class of PI3K pathway inhibitors that are uniquely cytotoxic to TNBC cells. This insight could be translated therapeutically by further developing Torin2 analogs or combinations of existing mTOR and PIKK inhibitors.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
(2′S)-2′-Deoxy-2′-fluoro-5-ethynyluridine, (F-ara-EdU), AldrichCPR
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Sodium pyrophosphate decahydrate, ACS reagent, ≥99%
Sigma-Aldrich
Torin2, ≥98% (HPLC)
Sigma-Aldrich
AZD6482, ≥98% (HPLC)
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
PIK93, ≥98% (HPLC)
Sigma-Aldrich
WYE-125132, ≥97% (HPLC)
Sigma-Aldrich
bpV(phen), A potent protein phosphotyrosine phosphatase inhibitor and insulin receptor kinase (IRK) activator.
Sigma-Aldrich
BX-912, ≥95% (HPLC)