Skip to Content
Merck
All Photos(1)

Key Documents

F1635

SAFC

Formaldehyde solution

contains 10-15% methanol as stabilizer, 37 wt. % in H2O

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.21
technique(s):
tissue processing: suitable
bp:
97 °C
vapor pressure:
52 mmHg ( 37 °C)

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

contains

10-15% methanol as stabilizer

concentration

37 wt. % in H2O
37-38%

technique(s)

tissue processing: suitable

impurities

≤0.03% free acid (as HCOOH)

color

APHA: ≤10

pH

2.8-4.0

bp

97 °C

mp

-15 °C

density

1.09 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is 37wt% solution of formaldehyde in water. It contains 10-15% methanol as stabilizer.

Application

Formaldehyde solution has been used to prepare formaldehyde fixation/permeabilization solution that is employed to fix tissue samples.

Biochem/physiol Actions

Induces tumors in the nasal epithelium of rats, and is suspected to be a human carcinogen. Potent mutagen.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

133.0 °F - closed cup

Flash Point(C)

56.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Omer Barkai et al.
Pain, 160(6), 1281-1296 (2019-04-02)
Endogenous inflammatory mediators contribute to the pathogenesis of pain by acting on nociceptors, specialized sensory neurons that detect noxious stimuli. Here, we describe a new factor mediating inflammatory pain. We show that platelet-derived growth factor (PDGF)-BB applied in vitro causes
Christopher Bruhn et al.
Genomics, proteomics & bioinformatics, 12(6), 255-265 (2014-12-03)
Cell cycle progression is coordinated with metabolism, signaling and other complex cellular functions. The investigation of cellular processes in a cell cycle stage-dependent manner is often the subject of modern molecular and cell biological research. Cell cycle synchronization and immunostaining
Cindy S Ma et al.
The Journal of experimental medicine, 205(7), 1551-1557 (2008-07-02)
Hyper-immunoglobulin E syndrome (HIES) is a primary immune deficiency characterized by abnormal and devastating susceptibility to a narrow spectrum of infections, most commonly Staphylococcus aureus and Candida albicans. Recent investigations have identified mutations in STAT3 in the majority of HIES
Arpeeta Sharma et al.
Diabetes, 64(11), 3937-3950 (2015-06-28)
Patients with diabetes have an increased risk of developing atherosclerosis. Endothelial dysfunction, characterized by the lowered bioavailability of endothelial NO synthase (eNOS)-derived NO, is a critical inducer of atherosclerosis. However, the protective aspect of eNOS in diabetes-associated atherosclerosis remains controversial
G R Moore et al.
Neurology, 55(10), 1506-1510 (2000-11-30)
To determine the pathologic basis of areas not exhibiting signal of the short-T2 component of the T2 relaxation distribution in MS, as studied in formalin-fixed brain. A myelin-specific MRI signal would be of great importance in assessing demyelination in patients

Articles

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service