Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pentane
Linear Formula:
CH
3
(CH
2
)
3
CH
3
CAS Number:
109-66-0
Molecular Weight:
72.15
EC Number:
203-692-4
Products
Product Category
solvents (7)
salts (2)
Brand
Sigma-Aldrich (10)
Supelco (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (2)
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
60489
Pentane,
≥99% (GC)
Expand
View Pricing
158941
Pentane,
reagent grade, 98%
Expand
View Pricing
34956
Pentane,
suitable for HPLC, ≥99.0%
Expand
View Pricing
76871
Pentane,
puriss. p.a., ≥99.0% (GC)
Expand
View Pricing
5.89576
Pentane,
≥99%, anhydrous
Expand
View Pricing
154954
Pentane,
spectrophotometric grade, ≥99%
Expand
View Pricing
236705
Pentane,
anhydrous, ≥99%
Expand
View Pricing
76870
Pentane,
analytical standard
Expand
View Pricing
1504933
Pentane,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PX0170
Pentane,
Expand
View Pricing
158941-M
Pentane,
reagent grade, 98%
Expand
View Pricing
34956-M
Pentane,
suitable for HPLC, ≥99.0%
Expand
View Pricing
60489-M
Pentane,
≥99% (GC)
Expand
View Pricing
154954-M
Pentane,
spectrophotometric grade, ≥99%
Expand
View Pricing