Skip to Content
Merck
All Photos(2)

Key Documents

154954

Sigma-Aldrich

Pentane

spectrophotometric grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.06
grade:
spectrophotometric grade
Assay:
≥99%
technique(s):
UV/Vis spectroscopy: suitable
bp:
35-36 °C (lit.)
vapor pressure:
26.98 psi ( 55 °C)
8.4 psi ( 20 °C)

grade

spectrophotometric grade

Quality Level

vapor density

2.48 (vs air)

vapor pressure

26.98 psi ( 55 °C)
8.4 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.010% water

evapn. residue

<0.0003%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 195 nm Amax: 1.00
λ: 200 nm Amax: 0.60
λ: 210 nm Amax: 0.30
λ: 220 nm Amax: 0.06
λ: 245-400 nm Amax: 0.01

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane (n-pentane) undergoes hydroisomerization in the presence of nanocrystalline beta (BEA) zeolite. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid. It isomerizes to iso-pentane in the presence of ZSM-5 (Zeolite Socony Mobil-5) zeolite catalysts under hydrogen and nitrogen atmosphere.
Pentane is a low boiling solvent, which finds applications in industries as a petroleum solvent and as a substitute for diethyl ether.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smallwood.McN I et al.
Solvent Recovery Handbook (2002)
Hydroisomerization of n-pentane over hybrid catalysts containing a supported hydrogenation catalyst.
Fujimoto K, et al.
Applied Catalysis A: General, 91(2), 81-86 (1992)
Fuming Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9174-9181 (2011-07-01)
Graphene oxide (GO) can be viewed as an amphiphilic soft material, which form thin films at organic solvent-water interfaces. However, organic solvent evaporation provides little driving force, which results in slow GO transfer in aqueous phase, thus dawdling GO film
Michael Polet et al.
Biomedical chromatography : BMC, 27(2), 259-266 (2012-07-21)
The administration of anabolic steroids is one of the most important issues in doping control and is detectable through a change in the carbon isotopic composition of testosterone and/or its metabolites. Gas chromatography-combustion-isotope ratio mass spectrometry (GC-C-IRMS), however, remains a
Dongsheng Guo et al.
Journal of hazardous materials, 186(2-3), 1788-1793 (2011-01-18)
The solvents n-pentane, methylene dichloride, ethyl ether and dodecylbenzenesulphonic acid sodium were used to regenerate exhausted activated carbon used in the process of treating coking wastewater, and the efficiency, ability, and optimum conditions of the different solvents on this regeneration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service