Skip to Content
Merck
All Photos(1)

Key Documents

154954-M

Supelco

Pentane

spectrophotometric grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.06
grade:
spectrophotometric grade
Assay:
≥99%
bp:
35-36 °C (lit.)

grade

spectrophotometric grade

vapor density

2.48 (vs air)

Assay

≥99%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

impurities

≤0.01% water

evapn. residue

≤0.0003%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

UV absorption

λ: 195 nm Amax: ≤1.00
λ: 200 nm Amax: ≤0.60
λ: 210 nm Amax: ≤0.30
λ: 220 nm Amax: ≤0.06
λ: 245 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.01

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service