Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
NanoFabTx
™
reagent
Products
Product Category
polymers (11)
stabilizers (4)
Brand
Sigma-Aldrich (15)
Physical Form
Quality Segment
Application
Polymer Composition
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
907766
NanoFabTx
™
reagent,
Stabilizer-Nano
Expand
View Pricing
911631
NanoFabTx
™
reagent,
Stabilizer-Micro
Expand
View Pricing
912808
NanoFabTx
™
reagent,
PEGPLGA-50L
Expand
View Pricing
916382
NanoFabTx
™
reagent,
PCL Nano
Expand
View Pricing
915955
NanoFabTx
™
reagent,
PEGPLGA-50H
Expand
View Pricing
912549
NanoFabTx
™
reagent,
PEGPCL-H
Expand
View Pricing
911305
NanoFabTx
™
reagent,
Stabilizer-F
Expand
View Pricing
919934
NanoFabTx
™
reagent,
PEGPLA-H-M
Expand
View Pricing
913049
NanoFabTx
™
reagent,
PEGPLGA-75L
Expand
View Pricing
915718
NanoFabTx
™
reagent,
PEGPLGA-75H
Expand
View Pricing
913308
NanoFabTx
™
reagent,
PEGPLA-L
Expand
View Pricing
919942
NanoFabTx
™
reagent,
PEGPLA-L-M
Expand
View Pricing
913448
NanoFabTx
™
reagent,
Stabilizer-P
Expand
View Pricing
915203
NanoFabTx
™
reagent,
PEGPCL-L
Expand
View Pricing
917788
NanoFabTx
™
reagent,
PEGPCL-UH
Expand
View Pricing