915955
NanoFabTx™ reagent
PEGPLGA-50H
Synonym(s):
Formulation kit, NanoFabTx™
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
NanoFabTx™ reagent, PEGPLGA-50H is a component of our NanoFabTx™ PEG-PLGA drug formulation screening kit (917796), which includes additional reagents and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles. NanoFabTx™ reagent, PEGPLGA-50H is a pegylated poly(lactic-co-glycolic acid) (PLGA) polymer. PLGA is a biocompatible and biodegradable polymer that has been used widely in drug delivery systems for controlled drug release of many different types of therapeutic molecules.
Application
NanoFabTx™ reagent, PEGPLGA-50H can be used as a refill for the component in our NanoFabTx™ PEG-PLGA drug formulation screening kit (917796).
Legal Information
NANOFABTX is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service