All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
Looking for similar products? Visit Product Comparison Guide
General description
NanoFabTx™ reagent, PEGPLGA-75L is a component of our NanoFabTx™ drug formulation screening kits (917796, 915408), which include additional reagents and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles. NanoFabTx™ reagent, PEGPLGA-75L is a pegylated poly(lactic-co-glycolic acid) (PLGA) polymer. PLGA is a biocompatible and biodegradable polymer that has been used widely in drug delivery systems for controlled drug release of many different types of therapeutic molecules.
Application
Legal Information
NANOFABTX is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service