Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Coated Wire Tube Rack
Products
Product Category
racks (19)
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
HS120088
Coated Wire Tube Rack,
to hold, 20 x 33 mm tubes, blue
Expand
View Pricing
HS120771
Coated Wire Tube Rack,
to hold, 24 x 35-40 mm tubes, blue
Expand
View Pricing
HS232461
Coated Wire Tube Rack,
to hold, 8 x 50 mL tubes, blue
Expand
View Pricing
HS120770
Coated Wire Tube Rack,
to hold, 48 x 25-30 mm tubes, blue
Expand
View Pricing
HS23108
Coated Wire Tube Rack,
to hold, 72 x 16 mm tubes, red
Expand
View Pricing
HS23072
Coated Wire Tube Rack,
to hold, 72 x 16 mm tubes, blue
Expand
View Pricing
HS120769
Coated Wire Tube Rack,
to hold, 24 x 20-25 mm tubes, blue
Expand
View Pricing
HS120087
Coated Wire Tube Rack,
to hold, 12 x 33 mm tubes, blue
Expand
View Pricing
HS120086
Coated Wire Tube Rack,
to hold, 20 x 25 mm tubes, blue
Expand
View Pricing
HS120089
Coated Wire Tube Rack,
to hold, 72 x 13 mm tubes, blue
Expand
View Pricing
HS120085
Coated Wire Tube Rack,
to hold, 12 x 25 mm tubes, blue
Expand
View Pricing
HS23215
Coated Wire Tube Rack,
to hold, 15 x 15 mL tubes, blue
Expand
View Pricing
HS120763
Coated Wire Tube Rack,
to hold, 108 x 13-16 mm tubes, blue
Expand
View Pricing
HS120765
Coated Wire Tube Rack,
to hold, 80 x 16-20 mm tubes, blue
Expand
View Pricing
HS120764
Coated Wire Tube Rack,
to hold, 40 x 16-20 mm tubes, blue
Expand
View Pricing
HS120772
Coated Wire Tube Rack,
to hold, 48 x 35-40 mm tubes, blue
Expand
View Pricing
HS120768
Coated Wire Tube Rack,
to hold, 80 x 20-25 mm tubes, blue
Expand
View Pricing
HS120767
Coated Wire Tube Rack,
to hold, 40 x 22-25 mm tubes, blue
Expand
View Pricing
HS120766
Coated Wire Tube Rack,
to hold, 40 x 18-22 mm tubes, blue
Expand
View Pricing