Key Documents
HS120763
Coated Wire Tube Rack
to hold, 108 x 13-16 mm tubes, blue
Synonym(s):
Tube holder, bottle rack
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
material
blue
high-density polyethylene coating
feature
to hold 108 x 13-16 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120763
L × W × H
9.3 in. (23.7 cm) × 5 in. (12.7 cm) × 2.6 in. (6.5 cm)
General description
The HDPE Coated Wire Rack is durable for transport or sample handling. It is chemically-resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.
Features and Benefits
  • Chemically-resistant HDPE Coated Wire Racks
  • Racks can be used between temperatures of -40°C and 121°C
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
  • AutoclavableMesh Bottom Support