Skip to Content
Merck
All Photos(1)

Key Documents

1570406

USP

1-Propanol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

autoignition temp.

700 °F

expl. lim.

13.7 %

manufacturer/tradename

USP

refractive index

n20/D 1.384 (lit.)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

1-Propanol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipankar Bhattacharya et al.
The Journal of general virology, 95(Pt 2), 363-372 (2013-09-03)
Non-structural 5A protein (NS5A) has emerged as an important pharmacological target for hepatitis C virus (HCV). However, little is known about the conformation of NS5A intracellularly or how NS5A inhibitors achieve the picomolar (pM) inhibition of virus replication. Here, we
Zak Perold et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 17(2), 190-206 (2014-06-18)
Pyrimethamine, an anti-malarial agent known to exhibit solid state polymorphism, may be purified by means of recrystallization. Recrystallization may alter the solid state chemistry of pharmaceuticals, which may impact the toxicity and/or manufacturability thereof. We evaluated the risks associated with
Sailyn Alfonso et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(14), 3660-3671 (2014-05-03)
Racotumomab-alum is an anti-idiotype vaccine targeting the NeuGcGM3 tumor-associated ganglioside. This clinical trial was conducted to provide a preliminary estimate of efficacy and safety of racotumomab as switch maintenance for patients with advanced non-small cell lung cancer (NSCLC). Patients with
Cornelia Neidlinger-Wilke et al.
Spine, 39(17), 1355-1364 (2014-05-17)
Conditioned media (CM) of cartilaginous endplates (CEPs) of intervertebral discs were analyzed in a bioassay with regard to their influence on matrix turnover and inflammatory factors on nucleus pulposus (NP) cells of the same patient. CEP tissue underwent further histological
Xiu-min Jiang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 957, 60-67 (2014-03-25)
In this study, a two-step process combining aqueous two-phase extraction (ATPE) with chromatography was developed for extraction and purification of alliin from garlic powder. The partition coefficient and yield value of alliin in different types of aqueous two-phase system (ATPS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service