Skip to Content
Merck
All Photos(3)

Key Documents

402893

Sigma-Aldrich

1-Propanol

1-Propanol

ACS reagent, ≥99.5%

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.05
grade:
ACS reagent
bp:
97 °C (lit.)
vapor pressure:
10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

dilution

(for analytical testing)

impurities

≤0.0004 meq/g Titr. acid
≤0.01% ethanol
≤0.01% methanol
≤0.03% Carbonyl compounds (as C2H5CHO)
≤0.05% CH3CHOHCH3
≤0.05% isopropanol
≤0.2% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

solubility

H2O: passes test
water: soluble

density

0.804 g/mL at 25 °C (lit.)

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-propanol is a primary alcohol that can be produced by oxidizing aliphatic hydrocarbons. It is extensively used as an organic solvent in the pharmaceutical, chemical, and food industries.

Application

1-Propanol can be used as a solvent in the:
  • Enzyme-catalyzed Henry-reaction of aromatic aldehydes and nitroalkanes to synthesize β-nitroalcohols.
  • Pd-catalyzed Suzuki-Miyaura cross-coupling reactions of aryl halides and arylboronic acids to synthesize biphenyl compounds.
  • Cross-coupling reaction of nitriles and amines to give secondary imines.
  • Synthesis of ZnO thin films by sol-gel method.
  • Extraction of brominated flame retardants from waste polymeric materials.
  • Synthesis of polyaniline/magnetic nanoparticles composite.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Exploration of the morphological transition phenomenon of polyaniline from microspheres to nanotubes in acid-free aqueous 1-propanol solution in a single polymerization process.
Huang YF and Lin CW.
Polymer International, 59(9), 1226-1232 (2010)
Vibrational spectra of liquid 1-propanol.
Michniewicz N, et al.
Journal of Molecular Structure, 887(1), 180-186 (2008)
Yu Deng et al.
Metabolic engineering, 13(5), 570-577 (2011-07-16)
Biofuel production from renewable resources can potentially address lots of social, economic and environmental issues but an efficient production method has yet to be established. Combinations of different starting materials, organisms and target fuels have been explored with the conversion
Growth of thin rubrene single crystals from 1-propanol solvent.
Matsukawa T, et al.
Journal of Crystal Growth, 312(2), 310-313 (2010)
Highly c-axis oriented ZnO thin film using 1-propanol as solvent in sol-gel synthesis.
Popa M, et al.
Materials Letters, 92, 267-270 (2013)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service