Skip to Content
Merck
All Photos(1)

Key Documents

1457301

USP

Naproxen

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Beilstein:
3591068
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

naproxen

manufacturer/tradename

USP

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

152-154 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1

InChI key

CMWTZPSULFXXJA-VIFPVBQESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naproxen USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dexamethasone Compounded Oral Suspension
  • Naproxen
  • Naproxen Compounded Oral Suspension
  • Naproxen Delayed-Release Tablets
  • Naproxen Oral Suspension
  • Naproxen Tablets
  • Terazosin Tablets

Biochem/physiol Actions

Non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Derry et al.
The Cochrane database of systematic reviews, (1)(1), CD004234-CD004234 (2009-01-23)
Naproxen, a non-steroidal anti-inflammatory drug, is used to treat various painful conditions including postoperative pain, and is often administered as the sodium salt to improve its solubility. This review updates a 2004 Cochrane review showing that naproxen sodium 550 mg
Marta L Capone et al.
Journal of the American College of Cardiology, 45(8), 1295-1301 (2005-04-20)
We investigated the occurrence of pharmacodynamic interaction between low-dose aspirin and naproxen. The uncertainty of cardioprotection by naproxen has encouraged its combination with aspirin in patients with arthritis and cardiovascular disease. The incubation of washed platelets with naproxen for 5
Chuthamanee C Suthisisang et al.
Headache, 50(5), 808-818 (2010-03-20)
To assess the efficacy and safety of naproxen sodium in the treatment of acute migraine attacks. Non-steroidal anti-inflammatory drugs including naproxen sodium have been used in treating migraine attack. A number of clinical trials of naproxen sodium in migraine have
Larisa Kovacevic et al.
Pediatric nephrology (Berlin, Germany), 18(8), 826-829 (2003-05-30)
A 17-year-old healthy girl was admitted to our hospital with diffuse abdominal pain and decreased oral intake of about 11 days duration. About a week prior to admission, she had taken naproxen, 250 mg four times a day for 4
Naoyoshi Nagata et al.
Journal of gastroenterology and hepatology, 30(2), 292-298 (2014-10-24)
The associations between antithrombotic or antihypertensive drugs and peptic ulcer bleeding (PUB) remain unknown, particularly in Asia, where Helicobacter pylori infection is prevalent. This study aims to evaluate the risks of PUB from antithrombotic drugs, angiotensin II receptor blockers (ARBs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service