Skip to Content
Merck
All Photos(1)

Key Documents

N0250000

Naproxen

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Beilstein:
3591068
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

naproxen

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

152-154 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1

InChI key

CMWTZPSULFXXJA-VIFPVBQESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naproxen EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Haupt et al.
Journal of biochemical and biophysical methods, 25(4), 273-284 (1992-12-01)
A column-switching system for fast determination of (R)- and (S)-naproxen in liver microsomes has been developed. The centrifuged sample was injected directly onto a pre-column with octadecylcoated silica. The retained analytes were then directed to an alpha 1-AGP column using
Michela Campolo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(11), 4489-4499 (2013-08-01)
Spinal cord trauma causes loss of motor function that is in part due to the ensuing inflammatory response. Hydrogen sulfide (H2S) is a potent, endogenous anti-inflammatory and neuroprotective substance that has been explored for use in the design of novel
Jenny-Maria Brozinski et al.
Environmental science & technology, 47(1), 342-348 (2012-11-29)
Pharmaceutical residues are ubiquitous in rivers, lakes, and at coastal waters affected by discharges from municipal wastewater treatment plants. In this study, the presence of 17 different pharmaceuticals and six different phase I metabolites was determined in the bile of
Jiayang Li et al.
Journal of the American Chemical Society, 135(2), 542-545 (2012-11-10)
As systemically used therapeutics for treating acute or chronic pains or inflammations, nonsteroidal anti-inflammatory drugs (NSAIDs) also associate with the adverse gastrointestinal and renal effects and cardiovascular risks. Thus, it is beneficial to develop topical gels that selectively inhibit cyclooxygenase-2
Dong Qing Zhang et al.
Chemosphere, 91(1), 14-21 (2012-12-27)
Scirpus validus was grown hydroponically and exposed to the pharmaceuticals, carbamazepine and naproxen at concentrations of 0.5-2.0 mg L(-1) for an exposure duration of up to 21 d. By the end of experiment, carbamazepine elimination from the nutrient solution reached

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service