Skip to Content
Merck
All Photos(1)

Key Documents

1270005

USP

Fentanyl citrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Fentanyl citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O · C6H8O7
CAS Number:
Molecular Weight:
528.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fentanyl

manufacturer/tradename

USP

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCC(=O)N(C1CCN(CC1)CCc2ccccc2)c3ccccc3

InChI

1S/C22H28N2O.C6H8O7/c1-2-22(25)24(20-11-7-4-8-12-20)21-14-17-23(18-15-21)16-13-19-9-5-3-6-10-19;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-12,21H,2,13-18H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

IVLVTNPOHDFFCJ-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fentanyl citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fentanyl Citrate Injection
  • Fentanyl Citrate Compounded Injection
  • Fentanyl Citrate and Bupivacaine Hydrochloride Compounded Injection
  • Fentanyl Citrate and Ropivacaine Hydrochloride Compounded Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fentanyl Citrate
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(5), 1848-1848 (2021)
Fentanyl Citrate and Ropivacaine Hydrochloride Compounded Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(1), 1851-1851 (2020)
Fentanyl Citrate Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(5), 1849-1849 (2021)
Fentanyl Citrate and Bupivacaine Hydrochloride Compounded Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(1), 1850-1850 (2020)
Fentanyl Citrate Compounded Injection
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(1), 1848-1848 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service