Skip to Content
Merck
All Photos(1)

Key Documents

415057

Sigma-Aldrich

Magnesium stearate

technical grade

Synonym(s):

Stearic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16CO2]2Mg
CAS Number:
Molecular Weight:
591.24
Beilstein:
3919702
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

powder

composition

palmitate salt, 25%
stearate salt, 65%

mp

200 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Mg]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Mg/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

HQKMJHAJHXVSDF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium stearate (Mg-St) is the magnesium salt of stearic acid. Its anhydrate, dihydrate and trihydrate forms have been prepared. The tabletting of the blends of magnesium stearate and lactose granules has been described. The influence of mixing time on hardness, disintegration time and ejection force on the compressed tablets was examined. Mg-St is widely used lubricant in pharmaceutical industry. It also plays a role in delaying the process of dissolution. Its detection in tablets by laser-induced breakdown spectroscopy has been proposed.

Magnesium stearate, the salt of stearic acid, is widely employed as an inactive component in making pharmaceutical tablets.

Application

Magnesium stearate may be used as excipient in the formulations of diclofenac sodium tablets. It may be used for the preparation of tramadol HCl matrix tablets.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical characterization of the polymorphic variations of magnesium stearate and magnesium palmitate hydrate species.
Sharpe SA, et al.
Structural Chemistry, 8(1), 73-84 (1997)
Magnesium stearate, a widely-used food additive, exhibits a lack of in vitro and in vivo genotoxic potential
Hobbs CA, et al.
Toxicology Reports, 4, 554-559 (2017)
Delay effect of magnesium stearate on tablet dissolution in acidic medium.
Ariyasu A, et al.
International Journal of Pharmaceutics, 511(2), 757-764 (2016)
Louis St-Onge et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 8(2), 272-288 (2005-08-30)
Laser-induced breakdown spectroscopy (LIBS) was evaluated for its potential as a process analytical tool for the rapid determination of magnesium stearate (MgSt) distribution within and between tablets as well as between batches in a typical manufacturing run, and for the
Effect of mixing time on the lubricating properties of magnesium stearate and the final characteristics of the compressed tablets.
Kikuta J-I and Kitamori N.
Drug Development and Industrial Pharmacy, 20(3), 343-355 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service